EMAIL THIS PAGE TO A FRIEND
Social work in health care
Perceptions of the hospital ethical environment among hospital social workers in the United States.
PMID 25760491
Abstract
Hospital social workers are in a unique context of practice, and one where the ethical environment has a profound influence on the ethical behavior. This study determined the ratings of ethical environment by hospital social workers in large nationwide sample. Correlates suggest by and compared to studies of ethical environment with nurses are explored. Positive ratings of the ethical environment are primarily associated with job satisfaction, as well as working in a centralized social work department and for a non-profit hospital. Religiosity and MSW education were not predictive. Implications and suggestions for managing the hospital ethical environment are provided.