EMAIL THIS PAGE TO A FRIEND
Molecular medicine reports
Estimation of the environmental effect of natural volatile organic compounds from Chamaecyparis obtusa and their effect on atopic dermatitis-like skin lesions in mice.
PMID 25760811
Abstract
Aromatherapy has been suggested as an alternative therapeutic method for the treatment of atopic dermatitis (AD), eczema and other skin diseases. In the current study, the anti-atopic properties of the volatile organic compounds of Chamaecyparis obtusa (VOCCo) were examined to determine whether they are amenable for use as a pharmaceutical candidate. The alterations in histological features, serum IgE levels and mast cell infiltration following exposure to VOCCo were determined in a 2,4-dinitrochlorobenzene (DNCB)-induced AD-like mouse model. The results of these experiments demonstrated that VOCCo inhibited the development of AD-like skin lesions by reducing the serum IgE level and mast cell infiltration into the dermal and subcutaneous layers. This was supported by screening of immune cytokine mRNAs, including interleukin (IL)-1β and IL-6 from the skin of DNCB-treated mice. The expression of IL-1β and IL-6 in the skin lesions of mice was dose-dependently inhibited by treatment with VOCCo. Furthermore, treatment with VOCCo resulted in the recovery of histopathological features in AD-like skin lesions. These results suggest that VOCCo may have therapeutic and preventive effects for the development of AD.