EMAIL THIS PAGE TO A FRIEND
Journal of health psychology
A narrative analysis of spiritual distress in geriatric physical rehabilitation.
PMID 25762383
Abstract
Drawing upon narrative data generated in a semi-structured interview with an 82-year-old female patient in geriatric physical rehabilitation, this clinical case study provides a detailed example of recognizing, assessing, and addressing spiritual distress as a symptom of physical pain. Data analysis focused on narrative content as well as on the interactive and performative aspects of narrating spiritual health issues in a close reading of two "attachment narratives." Results support the "narrative turn" in healthcare, including the therapeutic benefits of empathic listening as "narrative care" in geriatric rehabilitation and in healthcare in general.