EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Immune quiescence of the brain is set by astroglial connexin 43.
PMID 25762685
Abstract
In the normal brain, immune cell trafficking and immune responses are strictly controlled and limited. This unique homeostatic equilibrium, also called brain immune quiescence, is crucial to maintaining proper brain functions and is altered in various pathological processes, from chronic immunopathological disorders to cognitive and psychiatric impairments. To date, the precise nature of factors regulating the brain/immune system interrelationship is poorly understood. In the present study, we demonstrate that one of these regulating factors is Connexin 43 (Cx43), a gap junction protein highly expressed by astrocytes at the blood-brain barrier (BBB) interface. We show that, by setting the activated state of cerebral endothelium, astroglial Cx43 controls immune recruitment as well as antigen presentation mechanisms in the mouse brain. Consequently, in the absence of astroglial Cx43, recruited immune cells elaborate a specific humoral autoimmune response against the von Willebrand factor A domain-containing protein 5a, an extracellular matrix protein of the brain. Altogether, our results demonstrate that Cx43 is a new astroglial factor promoting the immune quiescence of the brain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32883
4-tert-Octylphenol monoethoxylate solution, 1 μg/mL in acetone, analytical standard
C16H26O2
32882
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
C16H26O2
14339
Bicinchoninic acid disodium salt hydrate, ≥99.0% (HPLC)
C20H10N2Na2O4 · xH2O
PHR1362
Calcium Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
C4H6CaO4
02672
Calcium atomic spectroscopy standard concentrate 1.00 g Ca, 1.00 g/L, for 1L standard solution, analytical standard
CaCl2
21059
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
CaCl2
21049
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
CaCl2
47829
D-(+) Glucose, analytical standard
C6H12O6
49152
D-(+)-Glucose, tested according to Ph.Eur.
C6H12O6
47249
D-(+)-Glucose, analytical standard
C6H12O6
PHR1000
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
C6H12O6
1181302
Dextrose, United States Pharmacopeia (USP) Reference Standard
C6H12O6
D9026
Digoxigenin, analytical standard
C23H34O5
Y0000640
Digoxigenin, European Pharmacopoeia (EP) Reference Standard
C23H34O5
63031
Magnesium ion solution for ISE, 0.01 M Mg, analytical standard (for ion-selective electrodes)
Cl2Mg
63042
Magnesium ion solution for ISE, 0.1 M Mg, analytical standard (for ion-selective electrodes)
Cl2Mg
S100
Mianserin hydrochloride, analytical standard, for drug analysis
C18H20N2 · HCl
84010
NBT Vials, for in vitro diagnostic use
C40H30Cl2N10O6
N6876
Nitrotetrazolium Blue chloride, ~98% (TLC)
C40H30Cl2N10O6
N6639
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
C40H30Cl2N10O6
74032
Nitrotetrazolium Blue chloride, ≥98.0% (calc. based on dry substance, T)
C40H30Cl2N10O6
792519
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
CHNaO3
1613655
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
CHNaO3
71631
Sodium bicarbonate, tested according to Ph.Eur.
CHNaO3
71383
Sodium chloride, tested according to Ph.Eur.
ClNa
71387
Sodium chloride, certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%
ClNa
PHR1321
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
ClNa
71177
Sodium standard for AAS, ready-to-use, traceable to BAM, in H2O
ClNa
84100
Sucrose, for microbiology, ACS reagent, ≥99.0%
C12H22O11
S1600000
Sucrose, European Pharmacopoeia (EP) Reference Standard
C12H22O11