EMAIL THIS PAGE TO A FRIEND
The Pediatric infectious disease journal
Prevalence and clinical burden of NDM-1 positive infections in pediatric and neonatal patients in Pakistan.
PMID 25764102
Abstract
In a neonatal/pediatric Gram-negative infection study from Islamabad, 71/82 strains were carbapenem resistant with 12/82 positive for New Delhi Metallo-β-lactamase and many being extensively antibiotic resistant. Burden and outcome analysis on 9 patients showed that 4/9 died after inadequate therapy regardless of organism type.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1392454
Meropenem, United States Pharmacopeia (USP) Reference Standard
C17H25N3O5S · 3H2O