EMAIL THIS PAGE TO A FRIEND
Food chemistry
Solubility and thermodynamic function of vanillin in ten different environmentally benign solvents.
PMID 25766824
Abstract
The solubility of vanillin in ten different environmentally benign solvents namely water, ethanol, ethylene glycol (EG), ethyl acetate (EA), isopropanol (IPA), propylene glycol (PG), polyethylene glycol-400 (PEG-400), Transcutol, butanol-1 and butanol-2 was measured and correlated at T=(298-318)K. The resulting experimental data were correlated with the modified Apelblat and Van't Hoff models. Both the models showed good correlation of experimental solubility data with calculated ones with root mean square deviations in the range of (0.08-1.55)%. The mole fraction solubility of vanillin was observed highest in PEG-400 (4.29 × 10(-1) at 298 K) followed by Transcutol, EA, butanol-2, ethanol, EG, PG, IPA, butanol-1 and water from T=(298-318)K. The results of thermodynamic function in terms of dissolution enthalpy, Gibbs energy and dissolution entropy showed endothermic, spontaneous and entropy-driven dissolution of vanillin in all environmentally benign solvents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
55417
1,2-Propanediol, analytical reference material
C3H8O2
12279
1,2-Propanediol, analytical standard
C3H8O2
82281
1,2-Propanediol, tested according to Ph.Eur.
C3H8O2
81780
1,3-Propanediol, puriss., ≥99.0% (GC)
C3H8O2
19422
1-Butanol, analytical standard
C4H10O
19420
1-Butanol, puriss. p.a., ACS reagent, ≥99.5% (GC)
C4H10O
PHR1209
1-Butanol, Pharmaceutical Secondary Standard; Certified Reference Material
C4H10O
294810
2-Butanol, anhydrous, 99.5%
C4H10O
96870
2-Butanol, analytical standard
C4H10O
PHR1563
2-Butanol, Pharmaceutical Secondary Standard; Certified Reference Material
C4H10O
19440
2-Butanol, puriss. p.a., Reag. Ph. Eur., ≥99.5% (GC)
C4H10O
B85919
2-Butanol, ReagentPlus®, ≥99%
C4H10O
77634
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
C8H8O3
Y0001547
Propylene glycol, European Pharmacopoeia (EP) Reference Standard
C3H8O2
1576708
Propylene glycol, United States Pharmacopeia (USP) Reference Standard
C3H8O2
PHR1051
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
C3H8O2
V2375
Vanillin
C8H8O3
30304
Vanillin, TraceCERT®, certified reference material
C8H8O3
94752
Vanillin, tested according to Ph.Eur.
C8H8O3
PHR1245
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
C8H8O3
V1104
Vanillin, ReagentPlus®, 99%
C8H8O3
W310700
Vanillin, ≥97%, FCC, FG
C8H8O3
W310727
Vanillin, natural, ≥97%, FCC, FG
C8H8O3
V0050000
Vanillin, European Pharmacopoeia (EP) Reference Standard
C8H8O3
PHR1091
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
C8H8O3
1711009
Vanillin Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
C8H8O3