EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Reductive stress impairs myoblasts mitochondrial function and triggers mitochondrial hormesis.
PMID 25769432
Abstract
Even though oxidative stress damage from excessive production of ROS is a well known phenomenon, the impact of reductive stress remains poorly understood. This study tested the hypothesis that cellular reductive stress could lead to mitochondrial malfunction, triggering a mitochondrial hormesis (mitohormesis) phenomenon able to protect mitochondria from the deleterious effects of statins. We performed several in vitro experiments on L6 myoblasts and studied the effects of N-acetylcysteine (NAC) at different exposure times. Direct NAC exposure (1mM) led to reductive stress, impairing mitochondrial function by decreasing maximal mitochondrial respiration and increasing H₂O₂production. After 24h of incubation, the reactive oxygen species (ROS) production was increased. The resulting mitochondrial oxidation activated mitochondrial biogenesis pathways at the mRNA level. After one week of exposure, mitochondria were well-adapted as shown by the decrease of cellular ROS, the increase of mitochondrial content, as well as of the antioxidant capacities. Atorvastatin (ATO) exposure (100μM) for 24h increased ROS levels, reduced the percentage of live cells, and increased the total percentage of apoptotic cells. NAC exposure during 3days failed to protect cells from the deleterious effects of statins. On the other hand, NAC pretreatment during one week triggered mitochondrial hormesis and reduced the deleterious effect of statins. These results contribute to a better understanding of the redox-dependant pathways linked to mitochondria, showing that reductive stress could trigger mitochondrial hormesis phenomenon.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3700853 Anti-Rabbit IgG (Fc specific), highly cross adsorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700863 Anti-Rabbit IgG (Fc specific)-Peroxidase antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700852 Anti-Rabbit IgG (Fc specific)-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700949 Anti-Rabbit IgG (H+L), F(ab′)2 fragment-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700978 Anti-Rabbit IgG (H+L), F(ab) fragment-Peroxidase antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700972 Anti-Rabbit IgG (H+L), F(ab) fragment-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700941 Anti-Rabbit IgG (H+L)-Peroxidase antibody produced in chicken, affinity isolated antibody, lyophilized powder
SAB3700934 Anti-Rabbit IgG (H+L)-Peroxidase antibody produced in donkey, affinity isolated antibody, lyophilized powder
SAB3700920 Anti-Rabbit IgG (H+L)-Peroxidase antibody produced in sheep, affinity isolated antibody, lyophilized powder
SAB3700842
Anti-Rabbit IgG F(ab′)2, F(ab′)2 fragment, highly cross absorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700832 Anti-Rabbit IgG F(ab′)2, highly cross adsorbed-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB3700831 Anti-Rabbit IgG F(ab′)2-Peroxidase antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB2501675
Anti-SOD2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501676
Anti-SOD2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406465
Anti-SOD2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2701618
Anti-sod2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701761
Anti-sod2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA001814
Anti-SOD2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102261
Anti-SOD2 antibody produced in rabbit, affinity isolated antibody
A7250
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
C5H9NO3S
A5099
N-Acetyl-L-cysteine, PharmaGrade, Ajinomoto, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production, suitable for cell culture
C5H9NO3S
A8199
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
C5H9NO3S
A9165
N-Acetyl-L-cysteine, cell culture tested, BioReagent
C5H9NO3S