EMAIL THIS PAGE TO A FRIEND
Clinical rheumatology
Rosuvastatin improves endothelial dysfunction in ankylosing spondylitis.
PMID 25771851
Abstract
Enhanced cardiovascular risk in ankylosing spondylitis (AS) provides a strong rationale for early therapeutical intervention. In view of the proven benefit of statins in atherosclerotic vascular disease, we aimed to investigate the effect of rosuvastatin on endothelial dysfunction (ED) and inflammatory disease activity in AS. In a single-blind, placebo-controlled, parallel study, 32 AS patients were randomized to receive 24xa0weeks of treatment with rosuvastatin (10xa0mg/day, n = 17) and placebo (n = 15) as an adjunct to existing stable antirheumatic drugs. Flow-mediated dilatation (FMD) was assessed by AngioDefender™ (Everest Genomic Ann Arbor, USA). Inflammatory measures (BASDAI, BASFI, CRP and ESR) and pro-inflammatory cytokines (tumour necrosis factor-alpha [TNF-α], interleukin-6 [IL-6] and interleukin-1 [IL-1]) were measured at baseline and after treatment. Lipids and adhesion molecules (intracellular adhesion molecule [ICAM-1] and vascular cell adhesion molecule [VCAM-1]) were estimated at baseline and after treatment. At baseline, inflammatory measures, pro inflammatory cytokines and adhesion molecules were elevated among both groups. After treatment with rosuvastatin, FMD improved significantly (p < 0.01). Levels of inflammatory measures, TNF-α, IL-6 and ICAM-1 decreased significantly (p < 0.01) after treatment with rosuvastatin. Rosuvastatin exerted positive effect on lipid spectrum. No significant change in the placebo group. Significant negative correlation was observed between FMD and IL-6, ICAM-1, CRP after treatment with rosuvastatin. First study to show that rosuvastatin improves inflammatory disease activity and ED in AS. Rosuvastatin lowers the proinflammatory cytokines, especially IL-6 and TNF-α, which downregulates adhesion molecules and CRP production which in turns improves ED. Improvement in ED in AS occurs through both cholesterol-independent and cholesterol-dependent pathways. Rosuvastatin can mediate modest but clinically apparent anti-inflammatory effects with modification of vascular risk factors in the context of high-grade autoimmune inflammation of AS.