EMAIL THIS PAGE TO A FRIEND
Gene
Haplotype combination of the bovine CFL2 gene sequence variants and association with growth traits in Qinchuan cattle.
PMID 25772133
Abstract
The aim of this study was to examine the association of cofilin2 (CFL2) gene polymorphisms with growth traits in Chinese Qinchuan cattle. Three single nucleotide polymorphisms (SNPs) were identified in the bovine CFL2 gene using DNA sequencing and (forced) PCR-RFLP methods. These polymorphisms included a missense mutation (NC_007319.5: g. C 2213 G) in exon 4, one synonymous mutation (NC_007319.5: g. T 1694 A) in exon 4, and a mutation (NC_007319.5: g. G 1500 A) in intron 2, respectively. In addition, we evaluated the haplotype frequency and linkage disequilibrium coefficient of three sequence variants in 488 individuals in QC cattle. All the three SNPs in QC cattle belonged to an intermediate level of genetic diversity (0.250.33). Association analysis indicated that SNP G 1500 A, T 1694 A and C 2213 G were significantly associated with growth traits in the QC population. The results of our study suggest that the CFL2 gene may be a strong candidate gene that affects growth traits in the QC cattle breeding program.