EMAIL THIS PAGE TO A FRIEND
Immunologic research
Immune protective effect of human alpha-1-antitrypsin gene during β cell transplantation in diabetic mice.
PMID 25772200
Abstract
Type 1 diabetes (T1D) is a chronic autoimmune disease in which β cells are destroyed. Islet transplantation is the most promising therapeutic treatment for T1D patients. However, allograft rejection and autoimmune reaction have been recognized as primary causes of graft loss after transplantation. Alpha-1-antitrypsin (AAT) is an important serine protease inhibitor in serum. AAT is characterized by anti-inflammation, anti-apoptosis, and induction-specific immunological tolerance. In this study, we successfully established NIT-hAAT cell lines, which are murine islet β cell lines with stable expression of human AAT (hAAT) gene. These NIT-hAAT cells were transplanted under the left kidney capsule of BALB/c diabetic mice. Interestingly, the sustained expression of hAAT in vivo can block the inflammatory cell infiltration and reduce the production of proinflammatory cytokines to effectively prevent nonspecific inflammation. Results showed that hAAT can inhibit the proliferation of lymphocytes, shift the balance between Th17 and Treg, and suppress the maturation of dendritic cells. Therefore, hAAT can serve as a beneficial immunomodulator that limits immune rejection to prolong islet allograft survival and achieve long-term successful transplant outcomes.