EMAIL THIS PAGE TO A FRIEND
Journal of neuroimmunology
Analysis of apoptosis-related genes in patients with clinically isolated syndrome and their association with conversion to multiple sclerosis.
PMID 25773154
Abstract
To analyse whether the expression of apoptotic transcripts is associated with the conversion from clinically isolated syndrome (CIS) to multiple sclerosis (MS). Eleven candidate transcripts belonging to the death receptor pathway, BCL-2, the inflammasome complex and NF-ΚB family were studied in the nonconverting and converting CIS patients during the four-year follow-up period. Conversion to MS was associated with marked variability in the expression of proapoptotic genes that were linked to TGF-B1 gene levels. The predominant expression of proapoptotic genes in patients with CIS suggests an increased potential to undergo apoptosis with the goal of terminating immune responses and regulating immune system homeostasis.