EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Submarine groundwater discharge as a major source of nutrients to the Mediterranean Sea.
PMID 25775554
Abstract
The Mediterranean Sea (MS) is a semienclosed basin that is considered one of the most oligotrophic seas in the world. In such an environment, inputs of allochthonous nutrients and micronutrients play an important role in sustaining primary productivity. Atmospheric deposition and riverine runoff have been traditionally considered the main external sources of nutrients to the MS, whereas the role of submarine groundwater discharge (SGD) has been largely ignored. However, given the large Mediterranean shore length relative to its surface area, SGD may be a major conveyor of dissolved compounds to the MS. Here, we used a (228)Ra mass balance to demonstrate that the total SGD contributes up to (0.3-4.8)⋅10(12) m(3) ⋅ y(-1) to the MS, which appears to be equal or larger by a factor of 16 to the riverine discharge. SGD is also a major source of dissolved inorganic nutrients to the MS, with median annual fluxes of 190⋅10(9), 0.7⋅10(9), and 110⋅10(9) mol for nitrogen, phosphorous, and silica, respectively, which are comparable to riverine and atmospheric inputs. This corroborates the profound implications that SGD may have for the biogeochemical cycles of the MS. Inputs of other dissolved compounds (e.g., iron, carbon) via SGD could also be significant and should be investigated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
294594
Mercury, electronic grade, 99.9999% trace metals basis
Hg