EMAIL THIS PAGE TO A FRIEND
Journal of bioscience and bioengineering
Anti-thrombotic effect of rutin isolated from Dendropanax morbifera Leveille.
PMID 25777266
Abstract
Dendropanax morbifera H. Lev. is well known in Korean traditional medicine for improvement of blood circulation. In this study, rutin, a bioflavonoid having anti-thrombotic and anticoagulant activities was isolated from a traditional medicinal plant, D. morbifera H. Lev. The chemical characteristics of rutin was studied to be quercetin 3-O-α-l-rhamnopyranosyl-(1-6)-β-d-glucopyranoside using high performance liquid chromatography mass spectrometry (HPLC-MS), proton nuclear magnetic resonance ((1)H NMR) and carbon-13 nuclear magnetic resonance ((13)C NMR). Turbidity and fibrin clotting studies revealed that rutin reduces fibrin clot in concentration dependent manner. Rutin was found to prolong activated partial thromboplastin time (aPTT), prothrombin time (PT) and closure time (CT). Furthermore, it decreased the activity of pro-coagulant protein, thrombin. In vivo study showed that rutin exerted a significant protective effect against collagen and epinephrine (or thrombin) induced acute thromboembolism in mice. These results suggest that rutin has a potent to be an anti-thrombotic agent for cardiovascular diseases.