EMAIL THIS PAGE TO A FRIEND
Journal of B.U.ON. : official journal of the Balkan Union of Oncology
67kDa laminin receptor regulates the activation of MAPKs through DUSPs in glioma cell line U251.
PMID 25778325
Abstract
All-trans-retinoic-acid (ATRA), the active derivative of vitamin A, is critical in regulating cell cycle as well as inhibiting tumor growth and angiogenesis. It has been used in the clinical treatment of leukemia. 67kDa laminin receptor (67LR), as one of the receptor of laminin, plays an important role in tumor cells invasion, proliferation and metastasis. Current research indicates that 67LR is highly expressed in glioma and is associated with tumor progression. However, the underlying molecular mechanisms, especially the signaling pathways involved, have not been reported yet. Therefore it is of great importance to clarify its downstream targets. The U251 glioma cell line was used in this study. Cell Counting Kit-8 was used in cell proliferation assay. Quantitative real-time PCR (qRT-PCR) was used to determine the transcription level of dual specificity phosphatases (DUSPs). Western blot analysis was used to detect the expression of mitogen activated protein kinases (MAPKs) and phosphorylated MAPKs. 67LR could influence the transcription of DUSPs and expression of MAPKs. ATRA could enhance the expression of 67LR in U251 cells and this enhancement was dose-dependent. ATRA was able to inhibit the growth of U251 cells. ATRA expressed significant therapeutic effect on glioma cells, and 67LR is not the only factor that can influence the proliferation of U251 cells.