EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Characterization of cyclic peptides containing disulfide bonds.
PMID 25778927
Abstract
Unlike linear peptides, analysis of cyclic peptides containing disulfide bonds is not straightforward and demands indirect methods to achieve a rigorous proof of structure. Three peptides that belong to this category, p-Cl-Phe-DPDPE, DPDPE, and CTOP, were analyzed and the results are presented in this paper. The great potential of two dimensional NMR and ESI tandem mass spectrometry was harnessed during the course of peptide characterizations. A new RP-HPLC method for the analysis of trifluoroacetic acid is also presented. It is robust, simple, and efficient compared to the currently available methods.