EMAIL THIS PAGE TO A FRIEND
The Journal of general virology
HIV-1 splicing is controlled by local RNA structure and binding of splicing regulatory proteins at the major 5' splice site.
PMID 25779589
Abstract
The 5' leader region of the human immunodeficiency virus 1 (HIV-1) RNA genome contains the major 5' splice site (ss) that is used in the production of the many spliced viral RNAs. This splice-donor (SD) region can fold into a stable stem-loop structure and the thermodynamic stability of this RNA hairpin influences splicing efficiency. In addition, splicing may be modulated by binding of splicing regulatory (SR) proteins, in particular SF2/ASF (SRSF1), SC35 (SRSF2), SRp40 (SRSF5) and SRp55 (SRSF6), to sequence elements in the SD region. The role of RNA structure and SR protein binding in splicing control was previously studied by functional analysis of mutant SD sequences. The interpretation of these studies was complicated by the fact that most mutations simultaneously affect both structure and sequence elements. We therefore tried to disentangle the contribution of these two variables by designing more precise SD region mutants with a single effect on either the sequence or the structure. The current analysis indicates that HIV-1 splicing at the major 5'ss is modulated by both the stability of the local RNA structure and the binding of splicing regulatory proteins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
117854
Trioctylphosphine, technical grade, 90%
C24H51P
718165
Trioctylphosphine, 97%
C24H51P