EMAIL THIS PAGE TO A FRIEND
Ulusal travma ve acil cerrahi dergisi = Turkish journal of trauma & emergency surgery : TJTES
Therapeutic evaluation of interleukin 1-beta antagonist Anakinra against traumatic brain injury in rats.
PMID 25779705
Abstract
The aim of this study was to evaluate the therapeutic efficiency of Anakinra, an IL-1ß antagonist with anti-inflammatory effects, in an experimental model of traumatic brain injury (TBI). Fifty-four rats underwent TBI after a weighted object was dropped onto a metal disc secured to their skulls. Animals were randomized into 3 main groups: control (n=18), TBI + saline (n=18; six animals per time-point) with samples obtained at the first, sixth and twenty-fourth h postoperatively, and TBI + Anakinra (n=18; six animals per time-point) with brain samples obtained at the first, sixth and twenty-fourth h postoperatively. Brain tissue and blood serum were extracted for the analysis of IL-1ß, malondialdehyde, glutathione peroxidase, superoxide dismutase, and catalase levels. Tissue sections were evaluated histopathologically under a light microscope. After trauma, tissue and serum IL-1ß levels were significantly elevated and after Anakinra administration, these levels substantially decreased. Glutathione peroxidase, superoxide dismutase, and catalase activity decreased following TBI and Anakinra administration proved effective in increasing the activity of these antioxidant enzymes. Histopathological analysis confirmed that Anakinra might protect the brain tissue and nerve cells from injury. Results demonstrate that Anakinra reduces the development of inflammation and tissue injury events associated with TBI.