EMAIL THIS PAGE TO A FRIEND
Reproductive sciences (Thousand Oaks, Calif.)
Mouse Ovarian Very Small Embryonic-Like Stem Cells Resist Chemotherapy and Retain Ability to Initiate Oocyte-Specific Differentiation.
PMID 25779995
Abstract
This study was undertaken to investigate stem cells in adult mouse ovary, the effect of chemotherapy on them and their potential to differentiate into germ cells. Very small embryonic-like stem cells (VSELs) that were SCA-1+/Lin-/CD45-, positive for nuclear octamer-binding transforming factor 4 (OCT-4), Nanog, and cell surface stage-specific embryonic antigen 1, were identified in adult mouse ovary. Chemotherapy resulted in complete loss of follicular reserve and cytoplasmic OCT-4 positive progenitors (ovarian germ stem cells) but VSELs survived. In ovarian surface epithelial (OSE) cell cultures from chemoablated ovary, proliferating germ cell clusters and mouse vasa homolog/growth differentiation factor 9-positive oocyte-like structure were observed by day 6, probably arising as a result of differentiation of the surviving VSELs. Follicle-stimulating hormone (FSH) exerted a direct stimulatory action on the OSE and induced stem cells proliferation and differentiation into premeiotic germ cell clusters during intact chemoablated ovaries culture. The FSH analog pregnant mare serum gonadotropin treatment to chemoablated mice increased the percentage of surviving VSELs in ovary. The results of this study provide evidence for the presence of potential VSELs in mouse ovaries and show that they survive chemotherapy, are modulated by FSH, and retain the ability to undergo oocyte-specific differentiation. These results show relevance to women who undergo premature ovarian failure because of oncotherapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4501173
Anti-Connexin 43 antibody produced in rabbit, affinity isolated antibody
SAB4501174
Anti-Connexin 43, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501175
Anti-Connexin 43, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1300064
Anti-GDF9 (N-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1405861
Anti-GDF9 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401149
Anti-GDF9 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4300504
Anti-GJA1 (Ab-367) antibody produced in rabbit, affinity isolated antibody
SAB2501246
Anti-GJA1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV36598
Anti-GJA1 antibody produced in rabbit, affinity isolated antibody
SAB2900237
Anti-GJA1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2100918
Anti-GJA1 antibody produced in rabbit, affinity isolated antibody
L1376
Linoleic acid, ≥99%
C18H32O2
L1012
Linoleic acid, liquid, BioReagent, suitable for cell culture
C18H32O2
L1268
Linoleic acid, ≥93% (GC)
C18H32O2
W338001
Linoleic acid, ≥95%
C18H32O2
62240
Linoleic acid, technical, 60-74% (GC)
C18H32O2
SAB4700284 Monoclonal Anti-CD15 antibody produced in mouse, clone MEM-158, purified immunoglobulin, buffered aqueous solution
WH0002697M1
Monoclonal Anti-GJA1 antibody produced in mouse, clone 3E5, purified immunoglobulin, buffered aqueous solution