EMAIL THIS PAGE TO A FRIEND
American journal of clinical pathology
Clinicopathologic evaluation of MYC expression in primary mediastinal (thymic) large B-cell lymphoma.
PMID 25780014
Abstract
Based on previous molecular studies, a small fraction of primary mediastinal (thymic) large B-cell lymphoma (PMBL) demonstrates MYC alterations. However, no studies have evaluated MYC protein expression by immunohistochemistry (IHC) with follow-up fluorescence in situ hybridization (FISH) analysis. We aim to evaluate the clinicopathologic importance of MYC IHC expression in PMBL. Three pathologists independently evaluated MYC IHC expression in 32 cases of PMBL for percent tumor positivity and nuclear intensity. FISH analysis for MYC rearrangement was performed on cases with high MYC IHC expression. Clinical data including treatment, follow-up, and outcome were also reviewed in a subset of cases. Variable MYC protein expression by IHC was detected in 30 (94%) of 32 cases of PMBL. One-third of the positive cases (10/30) showed high MYC IHC expression of at least 30% nuclear positivity. FISH analyses for MYC rearrangement on these 10 cases were negative. Review of clinical data on a subset of cases with high and low MYC IHC expression showed no differences in clinical outcome. MYC protein expression by IHC is present in most PMBLs. Increased MYC protein expression can be seen in one-third of the cases; however, it does not correlate with genetic abnormalities by FISH. There is also no significant impact of MYC protein expression on clinical outcomes.