EMAIL THIS PAGE TO A FRIEND
The Journal of clinical endocrinology and metabolism
Interchromosomal insertional translocation at Xq26.3 alters SOX3 expression in an individual with XX male sex reversal.
PMID 25781358
Abstract
46,XX male sex reversal occurs in approximately 1: 20 000 live births and is most commonly caused by interchromosomal translocations of the Y-linked sex-determining gene, SRY. Rearrangements of the closely related SOX3 gene on the X chromosome are also associated with 46,XX male sex reversal. It has been hypothesized that sex reversal in the latter is caused by ectopic expression of SOX3 in the developing urogenital ridge where it triggers male development by acting as an analog of SRY. However, altered regulation of SOX3 in individuals with XX male sex reversal has not been demonstrated. Here we report a boy with SRY-negative XX male sex reversal who was diagnosed at birth with a small phallus, mixed gonads, and borderline-normal T. Molecular characterization of the affected individual was performed using array comparative genomic hybridization, fluorescent in situ hybridization of metaphase chromosomes, whole-genome sequencing, and RT-PCR expression analysis of lymphoblast cell lines. The affected male carries ∼774-kb insertion translocation from chromosome 1 into a human-specific palindromic sequence 82 kb distal to SOX3. Importantly, robust SOX3 expression was identified in cells derived from the affected individual but not from control XX or XY cells, indicating that the translocation has a direct effect on SOX3 regulation. This is the first demonstration of altered SOX3 expression in an individual with XX male sex reversal and suggests that SOX3 can substitute for SRY to initiate male development in humans.