EMAIL THIS PAGE TO A FRIEND
Aging clinical and experimental research
Aging does not affect the ability of human monocyte-derived dendritic cells to phagocytose Candida albicans.
PMID 25783173
Abstract
Dendritic cells (DCs) are the most potent antigen-presenting cells, playing a key role in induction of both innate and adaptive immunity. Immunosenescence refers to age-associated changes in the immune system, which may be associated with susceptibility to infections and their clinical complications. The precise effects of aging on DCs in immunity to infections are not well understood. Among the common pathogenic microorganisms, the fungus Candida albicans is an important pathogen for the development of invasive infections, especially in immunocompromised individuals, as well as during aging. To make a comparative in vitro evaluation of the immunomodulatory function of DCs challenged with C. albicans, by phagocytosis of the fungal cells, and determine the involvement of TLR2 and TLR4 receptors. For this purpose, DCs were generated with the use of peripheral blood monocytes from healthy young and aged subjects. The phagocytosis of C. albicans is developed by DCs in TLR2- and TLR4-dependent way. This mechanism is not affected by aging. Given the important role of the DCs in responses against the fungus, it is evident that if changes in phagocytosis occurred with aging, impairment in the elderly could develop. However, the evidence that phagocytosis of this fungus by DCs is not impaired with aging, brings us to the question of which are the mechanisms truly associated with the prevalence of certain diseases in the elderly.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW22787 Anti-TLR2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
WH0007097M7
Monoclonal Anti-TLR2 antibody produced in mouse, clone 1E7, purified immunoglobulin, buffered aqueous solution
SAB1404474
Monoclonal Anti-TLR2 antibody produced in mouse, clone 2G9, purified immunoglobulin, buffered aqueous solution
SAB1409745
Monoclonal Anti-TLR2 antibody produced in mouse, clone 2A3, purified immunoglobulin, buffered aqueous solution
SAB1409746 Monoclonal Anti-TLR2 antibody produced in mouse, clone 2D2, purified immunoglobulin, buffered aqueous solution
SAB1409747 Monoclonal Anti-TLR2 antibody produced in mouse, clone 3F10, purified immunoglobulin, buffered aqueous solution
SAB1409748 Monoclonal Anti-TLR2 antibody produced in mouse, clone 1E3, purified immunoglobulin, buffered aqueous solution
SAB1409749 Monoclonal Anti-TLR2 antibody produced in mouse, clone 4D6, purified immunoglobulin, buffered aqueous solution
SAB1409750 Monoclonal Anti-TLR2 antibody produced in mouse, clone 4G9, purified immunoglobulin, buffered aqueous solution
SAB1409751 Monoclonal Anti-TLR2 antibody produced in mouse, clone 1C6, purified immunoglobulin, buffered aqueous solution
SAB1409752 Monoclonal Anti-TLR2 antibody produced in mouse, clone 1B10, purified immunoglobulin, buffered aqueous solution
SAB1409753 Monoclonal Anti-TLR2 antibody produced in mouse, clone 4C4, purified immunoglobulin, buffered aqueous solution
SAB1409754 Monoclonal Anti-TLR2 antibody produced in mouse, clone 3H8, purified immunoglobulin, buffered aqueous solution
SAB1409755 Monoclonal Anti-TLR2 antibody produced in mouse, clone 4H6, purified immunoglobulin, buffered aqueous solution