EMAIL THIS PAGE TO A FRIEND
Journal of virological methods
Analysis of protein expression changes of the Vero E6 cells infected with classic PEDV strain CV777 by using quantitative proteomic technique.
PMID 25783682
Abstract
Recent outbreaks of porcine epidemic diarrhea virus (PEDV) have caused widespread concern. The identification of proteins associated with PEDV infection might provide insight into PEDV pathogenesis and facilitate the development of novel antiviral strategies. We analyzed the differential protein profile of PEDV-infected Vero E6 cells using mass spectrometry and an isobaric tag for relative and absolute quantification. A total of 126 proteins were identified that were differentially expressed between the PEDV-infected and mock-infected groups (P<0.05, quantitative ratio ≥1.2), among which the expression of 58 proteins was up-regulated and that of 68 proteins was down-regulated in the PEDV-infected Vero E6 cells, involving in integrin β2/β3, cystatin-C. The Gene Ontology analysis indicated that the molecular function of the differentially expressed proteins (DEPs) was primarily related to binding and catalytic activity, and that the biological functions in which the DEPs are involved included metabolism, organismal systems, cellular processes, genetic information processing, environmental information processing, and diseases. Among the disease-related functions, certain anti-viral pathways and proteins, such as the RIG-I-like receptor, Rap1, autophagy, mitogen-activated protein kinase, PI3K-Akt and Jak-STAT signaling pathways, and integrin β2/β3 and cystatin-C proteins, represented potential factors in PEDV infection. Our findings provide valuable insight into PEDV-Vero E6 cell interactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA042215
Anti-BRI3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2501239
Anti-CST3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2700368
Anti-Cst3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA013143
Anti-CST3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401095
Anti-CST3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2500286
Anti-Cystatin 3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1411216
Anti-S100A2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1402739
Monoclonal Anti-CST3 antibody produced in mouse, clone 1H4, purified immunoglobulin, buffered aqueous solution
SAB4100452
Monoclonal Anti-CST3 antibody produced in mouse, clone CST3-A389, culture supernatant
SAB4100486
Monoclonal Anti-CST3 antibody produced in mouse, clone CST3-B389, culture supernatant
SAB4100533
Monoclonal Anti-CST3 antibody produced in mouse, clone CST3-A481, culture supernatant
SAB1408907 Monoclonal Anti-CST3 antibody produced in mouse, clone 2G4, purified immunoglobulin, buffered aqueous solution
SAB1408908
Monoclonal Anti-CST3 antibody produced in mouse, clone 3D10, purified immunoglobulin, buffered aqueous solution
S6797 Monoclonal Anti-S100A2 antibody produced in mouse, clone SH-L1, ascites fluid
SAB1404344
Monoclonal Anti-S100A2 antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous solution
SAB1409616
Monoclonal Anti-S100A2 antibody produced in mouse, clone 2D10-A3, purified immunoglobulin, buffered aqueous solution