EMAIL THIS PAGE TO A FRIEND
Molecular cancer research : MCR
Endophilin A2 Promotes TNBC Cell Invasion and Tumor Metastasis.
PMID 25784716
Abstract
Triple-negative breast cancers (TNBCs) are highly aggressive cancers that lack targeted therapies. However, EGFR is frequently activated in a subset of TNBCs and represents a viable clinical target. Because the endocytic adaptor protein Endophilin A2 (SH3GL1/Endo II) has been implicated in EGFR internalization, we investigated Endo II expression and function in human TNBCs. Endo II expression was high in several TNBC cells compared with normal breast epithelial cells. Stable knockdown (KD) of Endo II was achieved in two TNBC cell lines, and although cell viability was unaffected, defects in receptor-mediated endocytosis were observed. EGFR signaling to Erk and Akt kinases was impaired in Endo II KD cells, and this correlated with reduced rates of EGFR internalization and cell motility. Endo II KD cells also displayed defects in three dimensional (3D) cell invasion, and this correlated with impaired extracellular matrix degradation and internalization of MT1-MMP. Endo II silencing also caused a significant reduction in TNBC tumor growth and lung metastasis in mammary orthotopic tumor xenograft assays. In human breast tumor specimens, Endo II expression was highest in TNBC tumors compared with other subtypes, and at the level of gene expression, high Endo II was associated with reduced relapse-free survival in patients with basal-like breast cancers. Together, these results identify a positive role for Endo II in TNBC tumor metastasis and a potential link with poor prognosis. Endophilin A2 and related adaptor proteins represent important signaling hubs to target in metastatic cancers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4501901
Anti-MMP-14, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501902
Anti-MMP14 (Cleaved-Tyr112) antibody produced in rabbit, affinity isolated antibody
GW21125 Anti-MMP14 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB4504579
Anti-phospho-EGFR (pSer1026) antibody produced in rabbit, affinity isolated antibody
SAB4300062
Anti-phospho-EGFR (pSer1070) antibody produced in rabbit, affinity isolated antibody
SAB4504382
Anti-phospho-EGFR (pSer1070) antibody produced in rabbit, affinity isolated antibody
SAB4504580
Anti-phospho-EGFR (pSer1071) antibody produced in rabbit, affinity isolated antibody
SAB4504578
Anti-phospho-EGFR (pSer695) antibody produced in rabbit, affinity isolated antibody
SAB4300151
Anti-phospho-EGFR (pThr678) antibody produced in rabbit, affinity isolated antibody
SAB4503759
Anti-phospho-EGFR (pThr678) antibody produced in rabbit, affinity isolated antibody
SAB4300152
Anti-phospho-EGFR (pThr693) antibody produced in rabbit, affinity isolated antibody
SAB4503760
Anti-phospho-EGFR (pThr693) antibody produced in rabbit, affinity isolated antibody
SAB4503909
Anti-phospho-EGFR (pTyr1016) antibody produced in rabbit, affinity isolated antibody
SAB4504170
Anti-phospho-EGFR (pTyr1069) antibody produced in rabbit, affinity isolated antibody
SAB4300063
Anti-phospho-EGFR (pTyr1092) antibody produced in rabbit, affinity isolated antibody
SAB4504383
Anti-phospho-EGFR (pTyr1092) antibody produced in rabbit, affinity isolated antibody
SAB4300209
Anti-phospho-EGFR (pTyr1110) antibody produced in rabbit, affinity isolated antibody
SAB4504384
Anti-phospho-EGFR (pTyr1110) antibody produced in rabbit, affinity isolated antibody
SAB4300177
Anti-phospho-EGFR (pTyr1172) antibody produced in rabbit, affinity isolated antibody
SAB4504385
Anti-phospho-EGFR (pTyr1172) antibody produced in rabbit, affinity isolated antibody
SAB4300285
Anti-phospho-EGFR (pTyr1197) antibody produced in goat, affinity isolated antibody
SAB4300183
Anti-phospho-EGFR (pTyr1197) antibody produced in rabbit, affinity isolated antibody
SAB4504386
Anti-phospho-EGFR (pTyr1197) antibody produced in rabbit, affinity isolated antibody
SAB4300184
Anti-phospho-EGFR (pTyr869) antibody produced in rabbit, affinity isolated antibody
SAB4504387
Anti-phospho-EGFR (pTyr869) antibody produced in rabbit, affinity isolated antibody
E6154 Anti-phospho-Epidermal Growth Factor Receptor (pTyr1068) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
E1530 Anti-phospho-Epidermal Growth Factor Receptor (pTyr1086) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
E6279 Anti-phospho-Epidermal Growth Factor Receptor (pTyr1173) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
E6029 Anti-phospho-Epidermal Growth Factor Receptor (pTyr845) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution
E0907 Anti-phospho-Epidermal Growth Factor Receptor (pTyr845) antibody produced in rabbit, affinity isolated antibody, lyophilized powder