EMAIL THIS PAGE TO A FRIEND
Small (Weinheim an der Bergstrasse, Germany)
Caterpillar-Inspired Design and Fabrication of A Self-Walking Actuator with Anisotropy, Gradient, and Instant Response.
PMID 25787120
Abstract
A caterpillar-mimetic bilayer actuator is reported, based on a wrinkled polydimethylsiloxane elastomer decorated with a hydroresponsive polyelectrolyte brush. The actuator can fold ultrafast into complex three-dimensional structures upon a change in relative humidity of the surrounding air. The folding direction is determined by the geometry of the actuator, the orientation of the wrinkles, and the gradient in polymer height on the surface. And some unique structures such as helicoid and gradient can be obtained that are seldom reported in the case of bilayered devices. The adaptive bending movement is very fast, anisotropic, highly efficient, and reversible. When the environmental humidity is repeatedly cycled up and down, the small device walks on a surface with a roughness gradient, which is reminiscent of the muscle-like movements.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
251658
3-Sulfopropyl methacrylate potassium salt, 98%
C7H11KO5S