EMAIL THIS PAGE TO A FRIEND
Cancer immunology research
CD4(+) T-Helper Type 1 Cytokines and Trastuzumab Facilitate CD8(+) T-cell Targeting of HER2/neu-Expressing Cancers.
PMID 25791067
Abstract
Vaccination strategies incorporating the immunodominant HLA-A2-restricted HER2/neu-derived peptide 369-377 (HER2369-377) are increasingly utilized in HER2/neu-expressing cancer patients. The failure of postvaccination HER2369-377-specific CD8(+) T cells to recognize HLA-A2(pos)HER2/neu-expressing cells in vitro, however, has been attributed to impaired MHC class I/HLA-A2 presentation observed in HER2/neu-overexpressing tumors. We reconcile this controversy by demonstrating that HER2369-377 is directly recognized by high functional-avidity HER2369-377-specific CD8(+) T cells-either genetically modified to express a novel HER2369-377 TCR or sensitized using HER2369-377-pulsed type 1-polarized dendritic cells (DC1)-on class I-abundant HER2(low), but not class I-deficient HER2(high), cancer cells. Importantly, a critical cooperation between CD4(+) T-helper type-1 (Th1) cytokines IFNγ/TNFα and HER2/neu-targeted antibody trastuzumab is necessary to restore class I expression in HER2(high) cancers, thereby facilitating recognition and lysis of these cells by HER2369-377-specific CD8(+) T cells. Concomitant induction of PD-L1 on HER2/neu-expressing cells by IFNγ/TNF and trastuzumab, however, has minimal impact on DC1-sensitized HER2369-377-CD8(+) T-cell-mediated cytotoxicity. Although activation of EGFR and HER3 signaling significantly abrogates IFNγ/TNFα and trastuzumab-induced class I restoration, EGFR/HER3 receptor blockade rescues class I expression and ensuing HER2369-377-CD8(+) cytotoxicity of HER2/neu-expressing cells. Thus, combinations of CD4(+) Th1 immune interventions and multivalent targeting of HER family members may be required for optimal anti-HER2/neu CD8(+) T-cell-directed immunotherapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300352
Anti-EGFR (Ab-1070) antibody produced in rabbit, affinity isolated antibody
SAB4300353
Anti-EGFR (Ab-1092) antibody produced in rabbit, affinity isolated antibody
SAB4300510
Anti-EGFR (Ab-1110) antibody produced in rabbit, affinity isolated antibody
SAB4300472
Anti-EGFR (Ab-1172) antibody produced in rabbit, affinity isolated antibody
SAB4300602
Anti-EGFR (Ab-1197) antibody produced in goat, affinity isolated antibody
SAB4300480
Anti-EGFR (Ab-1197) antibody produced in rabbit, affinity isolated antibody
SAB4300453
Anti-EGFR (Ab-678) antibody produced in rabbit, affinity isolated antibody
SAB4300454
Anti-EGFR (Ab-693) antibody produced in rabbit, affinity isolated antibody
SAB4300481
Anti-EGFR (Ab-869) antibody produced in rabbit, affinity isolated antibody
GW21146 Anti-EGFR antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1405746
Anti-EGFR antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA001200
Anti-EGFR antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA018530
Anti-EGFR antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500711
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
SAB4500712
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
SAB4500720
Anti-EGFR antibody produced in rabbit, affinity isolated antibody
SAB4500713
Anti-EGFR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500714
Anti-EGFR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500715
Anti-EGFR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500716
Anti-EGFR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500717
Anti-EGFR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500718
Anti-EGFR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500719
Anti-EGFR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500721
Anti-EGFR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500722
Anti-EGFR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4500723
Anti-EGFR, C-Terminal antibody produced in rabbit, affinity isolated antibody
E1282 Anti-Epidermal Growth Factor Receptor antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB4300579
Anti-ERBB3 (Ab-1289) antibody produced in rabbit, affinity isolated antibody
SAB4300580
Anti-ERBB3 (Ab-1328) antibody produced in rabbit, affinity isolated antibody
HPA045396
Anti-ERBB3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution