EMAIL THIS PAGE TO A FRIEND
Acta histochemica
Different pediatric brain tumors are associated with different gene expression profiling.
PMID 25792036
Abstract
Malignant brain tumors are the most common pediatric solid tumors and are the leading cause of death from childhood cancers. These tumors include several histologic subtypes. Due to the particular properties of brain tumors, such as growth and division, examination of brain tumors and the analysis of results are not simple. Up to date there is a dearth of useful biomarkers that have been validated and clinically implemented for pediatric brain tumors. In order to identify the new genetic alterations we recognized, using microarray dataset, chitinases as new potential biomarkers of CNS tumors. The modulation of chitinases was confirmed also in the different histologic subtypes. Our study revealed that distinct patterns of chitinases expression characterize the diverse histological subtypes. In addition evaluating other lisosomal enzymes such as glycosidases and proteases we found that NEU4, CTBS and GBA2 belonging to glycosidases family and CTSC, CTSK and CTSF belonging to proteases family were differently modulated. Future investigations are needed to be performed before some of these enzymes could finally be used as biomarkers of specific types of CNS neoplasms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405615
Anti-CHI3L2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA005443
Anti-CHI3L2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2700845
Anti-CHI3L2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701165
Anti-CHI3L2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1400499
Anti-CHIA antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB2100419
Anti-CHIA antibody produced in rabbit, affinity isolated antibody
HPA059193
Anti-CHIA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410534
Anti-CHIA antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2103322
Anti-CHIA, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1104214
Anti-CHID1 (126-140) antibody produced in rabbit, IgG fraction of antiserum
SAB1304440
ANTI-CHID1 (C-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA039374
Anti-CHID1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2501721
Anti-CHIT1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1401072
Anti-CHIT1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA010575
Anti-CHIT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401073
Anti-CHIT1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1402723
Monoclonal Anti-CHIT1 antibody produced in mouse, clone 1D11, purified immunoglobulin, buffered aqueous solution
SAB5300323 Monoclonal Anti-CHIT1 antibody produced in mouse, clone 1D9G2, ascites fluid