EMAIL THIS PAGE TO A FRIEND
Human genetics
Homozygous MED25 mutation implicated in eye-intellectual disability syndrome.
PMID 25792360
Abstract
Genetic syndromes involving both brain and eye abnormalities are numerous and include syndromes such as Warburg micro syndrome, Kaufman oculocerebrofacial syndrome, Cerebro-oculo-facio-skeletal syndrome, Kahrizi syndrome and others. Using exome sequencing, we have been able to identify homozygous mutation p.(Tyr39Cys) in MED25 as the cause of a syndrome characterized by eye, brain, cardiac and palatal abnormalities as well as growth retardation, microcephaly and severe intellectual disability in seven patients from four unrelated families, all originating from the same village. The protein encoded by MED25 belongs to Mediator complex or MED complex, which is an evolutionary conserved multi-subunit RNA polymerase II transcriptional regulator complex. The MED25 point mutation is located in the von Willebrand factor type A (MED25 VWA) domain which is responsible for MED25 recruitment into the Mediator complex; co-immunoprecipitation experiment demonstrated that this mutation dramatically impairs MED25 interaction with the Mediator complex in mammalian cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1411939
ANTI-CDK8 antibody produced in mouse, clone 5H4, purified immunoglobulin, buffered aqueous solution
SAB1411940
ANTI-CDK8 antibody produced in mouse, clone 2E6, purified immunoglobulin, buffered aqueous solution
AV03050
Anti-CDK8 antibody produced in rabbit, IgG fraction of antiserum
M9817
Anti-MBD4 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
HPA002031
Anti-MBD4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101442
Anti-MBD4 antibody produced in rabbit, affinity isolated antibody
SAB4502265
Anti-MED1 antibody produced in rabbit, affinity isolated antibody
AV31452
Anti-MED16 antibody produced in rabbit, affinity isolated antibody
SAB2101454
Anti-MED16 antibody produced in rabbit, affinity isolated antibody
HPA030685
Anti-MED16 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104647
Anti-MED23, (N-terminal) antibody produced in rabbit, affinity isolated antibody
HPA039547
Anti-MED24 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503717
Anti-MED24, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1406909
Anti-MED6 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2101458
Anti-MED6 antibody produced in rabbit, affinity isolated antibody
SAB2103398
Anti-MED6 antibody produced in rabbit, affinity isolated antibody
SAB1410053
Anti-MED6 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1411807
ANTI-PPARBP antibody produced in mouse, clone 1G4, purified immunoglobulin, buffered aqueous solution
SAB1411808
ANTI-PPARBP antibody produced in mouse, clone 2G1, purified immunoglobulin, buffered aqueous solution
SAB1411809
ANTI-PPARBP antibody produced in mouse, clone 2H6, purified immunoglobulin, buffered aqueous solution
SAB1411810
ANTI-PPARBP antibody produced in mouse, clone 3F6, purified immunoglobulin, buffered aqueous solution
SAB1411881
ANTI-THRAP5 antibody produced in mouse, clone 3B10, purified immunoglobulin, buffered aqueous solution
SAB1411886
ANTI-THRAP5 antibody produced in mouse, clone 3A3, purified immunoglobulin, buffered aqueous solution
AV32301
Anti-THRAP5 antibody produced in rabbit, IgG fraction of antiserum
WH0001024M1
Monoclonal Anti-CDK8 antibody produced in mouse, clone 6H5, purified immunoglobulin, buffered aqueous solution
SAB1403658
Monoclonal Anti-CDK8, (C-terminal) antibody produced in mouse, clone 6E5, purified immunoglobulin, buffered aqueous solution
SAB1403024
Monoclonal Anti-CRSP3 antibody produced in mouse, clone 4H6, purified immunoglobulin, buffered aqueous solution
SAB1400371
Monoclonal Anti-MED6 antibody produced in mouse, clone 4C2, purified immunoglobulin, buffered aqueous solution
WH0005469M1
Monoclonal Anti-PPARBP antibody produced in mouse, clone 2A2, purified immunoglobulin, buffered aqueous solution
SAB1404231
Monoclonal Anti-PPARBP antibody produced in mouse, clone 1G3, purified immunoglobulin, buffered aqueous solution