EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
The calcium-binding proteins S100A8 and S100A9 initiate the early inflammatory program in injured peripheral nerves.
PMID 25792748
Abstract
To shed light on the early immune response processes in severed peripheral nerves, we performed genome-wide transcriptional profiling and bioinformatics analyses of the proximal (P, regenerating) and distal (D, degenerating) nerve stumps on day 1 in the sciatic nerve axotomy model in rats. Multiple cell death-related pathways were activated in the degenerating D stump, whereas activation of the cytoskeletal motility and gluconeogenesis/glycolysis pathways was most prominent in the P stump of the axotomized nerve. Our bioinformatics analyses also identified the specific immunomodulatory genes of the chemokine, IL, TNF, MHC, immunoglobulin-binding Fc receptor, calcium-binding S100, matrix metalloproteinase, tissue inhibitor of metalloproteinase, and ion channel families affected in both the P and D segments. S100a8 and S100a9 were the top up-regulated genes in both the P and D segments. Stimulation of cultured Schwann cells using the purified S100A8/A9 heterodimer recapitulated activation of the myeloid cell and phagocyte chemotactic genes and pathways, which we initially observed in injured nerves. S100A8/A9 heterodimer injection into the intact nerve stimulated macrophage infiltration. We conclude that, following peripheral nerve injury, an immediate acute immune response occurs both distal and proximal to the lesion site and that the rapid transcriptional activation of the S100a8 and S100a9 genes results in S100A8/A9 hetero- and homodimers, which stimulate the release of chemokines and cytokines by activated Schwann cells and generate the initial chemotactic gradient that guides the transmigration of hematogenous immune cells into the injured nerve.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
82320
(±)-Propylene oxide, puriss. p.a., ≥99.5% (GC)
C3H6O
110205
(±)-Propylene oxide, ReagentPlus®, ≥99%
C3H6O
471968
(±)-Propylene oxide, ReagentPlus®, 99%
C3H6O
SAB1401062
Anti-CD68 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2700244
Anti-CD68 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA048982
Anti-CD68 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107746
Anti-CD68 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
SAB2107747
Anti-CD68 antibody produced in rabbit, affinity isolated antibody
SAB2500903
Anti-S100A9 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA004193
Anti-S100A9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411220
Anti-S100A9 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2104737
Anti-S100A9, (N-terminal) antibody produced in rabbit, affinity isolated antibody
HPA015768
Anti-S100B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106315
Anti-S100B antibody produced in rabbit, affinity isolated antibody, lyophilized powder
D0899
Diazepam
C16H13ClN2O
D-907
Diazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material
C16H13ClN2O
F6886
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
C22H34O7
F3917
Forskolin, For use in molecular biology applications
C22H34O7
M8884
Methyl Green, zinc chloride salt, ~85%
C27H35BrClN3 · ZnCl2
67060
Methyl Green, zinc chloride salt, for microscopy (Bact., Bot., Hist.)
C27H35BrClN3 · ZnCl2
198080
Methyl Green, zinc chloride salt, certified by the Biological Stain Commission, Dye content 85 %
C27H35BrClN3 · ZnCl2
323829
Methyl Green, zinc chloride salt, <0.5% crystal violet, Dye content 80 %
C27H35BrClN3 · ZnCl2
AMAB90873
Monoclonal Anti-CD68 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1338, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90874
Monoclonal Anti-CD68 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1346, purified immunoglobulin, buffered aqueous glycerol solution
S2532
Monoclonal Anti-S-100 (β-Subunit) antibody produced in mouse, clone SH-B1, ascites fluid
SAB1404348
Monoclonal Anti-S100A9 antibody produced in mouse, clone 4G9, purified immunoglobulin, buffered aqueous solution
SAB1409623
Monoclonal Anti-S100A9 antibody produced in mouse, clone 1C22, purified immunoglobulin, buffered aqueous solution
239690
Succinic anhydride, ≥99% (GC)
C4H4O3