EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Peptide/protein stapling and unstapling: introduction of s-tetrazine, photochemical release, and regeneration of the peptide/protein.
PMID 25793939
Abstract
Protocols have been achieved that permit facile introduction of s-tetrazine into unprotected peptides and the protein, thioredoxin, between two cysteine sulfhydryl groups (i.e., staple), followed by photochemical release (i.e., unstaple) and regeneration of the peptide/protein upon removal of the cyano groups from the derived bisthiocyanate. The S,S-tetrazine macrocycles in turn provide a convenient handle for probe introduction by exploiting the inverse electron demand Diels-Alder reactivity of the tetrazine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
792594
3,6-Dichloro-1,2,4,5-tetrazine, 96%
C2Cl2N4