EMAIL THIS PAGE TO A FRIEND
Journal of affective disorders
Risk of developing major depressive disorder and anxiety disorders among adolescents and adults with atopic dermatitis: a nationwide longitudinal study.
PMID 25795537
Abstract
Previous cross-sectional studies have suggested a comorbid association between atopic dermatitis (AD) and depressive disorder as well as anxiety disorders, but the temporal relationship was not determined. Using the Taiwan National Health Insurance Research Database, 8208 AD patients aged 12 and older without psychiatric history and age-/sex-matched (1:1) controls between 1998 and 2008 were enrolled in our study and followed to the end of 2011. Subjects who developed major depression, any depressive disorder, and anxiety disorders during the follow-up were identified. The Cox regression analysis after adjusting for demographic data and atopic comorbidities demonstrated that patients with AD had an elevated risk of developing major depression (hazard ratio [HR]: 6.56, 95% confidence interval [CI]: 3.64-11.84), any depressive disorder (HR: 5.44, 95% CI: 3.99-7.44), and anxiety disorders (HR: 3.57, 95% CI: 2.55-4.98). Stratified by age group, both adolescents and adults with AD were prone to developing major depression (HR: 4.26, 95% CI: 1.39-13.13; HR: 7.56, 95% CI: 3.75-15.23), any depressive disorder (HR: 4.38, 95% CI: 2.09-9.18; HR: 5.66, 95% CI: 4.01-7.99), and anxiety disorders (HR: 5.40, 95% CI: 2.02-14.39; HR: 3.36, 95% CI: 2.38-4.80). AD in both adolescence and adulthood increased the risk of developing major depression, any depressive disorder, and anxiety disorders in later life. Further studies would be required to clarify the possible underlying mechanism between AD and depression as well as anxiety disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
713236
(9R)-6′-Methoxycinchonan-9-amine trihydrochloride, ≥90.0%
C20H25N3O · 3HCl