EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Caspase-1-independent IL-1 release mediates blister formation in autoantibody-induced tissue injury through modulation of endothelial adhesion molecules.
PMID 25795756
Abstract
Although reports documented aberrant cytokine expression in autoimmune bullous dermatoses (AIBDs), cytokine-targeting therapies have not been established in these disorders. We showed previously that IL-6 treatment protected against tissue destruction in experimental epidermolysis bullosa acquisita (EBA), an AIBD caused by autoantibodies to type VII collagen (COL7). The anti-inflammatory effects of IL-6 were mediated by induction of IL-1ra, and prophylactic IL-1ra administration prevented blistering. In this article, we demonstrate elevated serum concentrations of IL-1β in both mice with experimental EBA induced by injection of anti-COL7 IgG and in EBA patients. Increased IL-1α and IL-1β expression also was observed in the skin of anti-COL7 IgG-injected wild-type mice compared with the significantly less diseased IL-1R-deficient or wild-type mice treated with the IL-1R antagonist anakinra or anti-IL-1β. These findings suggested that IL-1 contributed to recruitment of inflammatory cells into the skin. Accordingly, the expression of ICAM-1 was decreased in IL-1R-deficient and anakinra-treated mice injected with anti-COL7. This effect appeared to be specifically attributable to IL-1 because anakinra blocked the upregulation of different endothelial adhesion molecules on IL-1-stimulated, but not on TNF-α-stimulated, cultured endothelial cells. Interestingly, injection of caspase-1/11-deficient mice with anti-COL7 IgG led to the same extent of skin lesions as in wild-type mice. Collectively, our data suggest that IL-1, independently of caspase-1, contributes to the pathogenesis of EBA. Because anti-IL-1β in a prophylactic setting and anakinra in a quasi-therapeutic setting (i.e., when skin lesions had already developed) improved experimental EBA, IL-1 appears to be a potential therapeutic target for EBA and related AIBDs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4501486
Anti-ICAM-1, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4300383
Anti-ICAM1 (Ab-512) antibody produced in rabbit, affinity isolated antibody
SAB2502120
Anti-ICAM1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1405977
Anti-ICAM1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA002126
Anti-ICAM1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA004877
Anti-ICAM1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406422
Anti-SELE antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1411245
Anti-SELE antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4700326 Monoclonal Anti-CD54 antibody produced in mouse, clone MEM-112, purified immunoglobulin, buffered aqueous solution
SAB4700327
Monoclonal Anti-CD54 antibody produced in mouse, clone MEM-111, purified immunoglobulin, buffered aqueous solution
SAB4700571 Monoclonal Anti-CD54 antibody produced in rat, clone YN1/1.7.4, purified immunoglobulin, buffered aqueous solution
WH0003383M1
Monoclonal Anti-ICAM1 antibody produced in mouse, clone 3H8-2G6, purified immunoglobulin, buffered aqueous solution