EMAIL THIS PAGE TO A FRIEND
The Journal of surgical research
Pediatric vascular injury: experience of a level 1 trauma center.
PMID 25796108
Abstract
Our purpose was to analyze modern major vascular injury (MVI) patterns in pediatric trauma, interventions performed, and outcomes at a level 1 trauma center. From January 2000-December 2012, all pediatric admissions (≤17 y) were reviewed. Of 1928 pediatric admissions, 103 (5.3%) sustained MVI. This cohort was 85% male, age 15 ± 3 y, 55% black, 58% penetrating, injury severity score of 23 ± 15, with a length of stay of 8 (5) days. Firearm-related injury (47%) was the most common mechanism. Location of injuries included the extremities (50.5%), abdomen/pelvis (29.1%), and chest/neck (20.4%). Operative procedures included repair/bypass (71.4%), ligation (12.4%), amputation (10.5%), or temporary shunt (2.9%). Only three injuries (2.9%) were treated endovascularly. MVI patients had a mortality rate of 19.4%, higher than the overall pediatric trauma population of 3.5% (P < 0.001). After logistic regression, independent risk factors of mortality were vascular injury to the neck (odds ratio [OR]: 6.5; confidence interval (CI): 1.1-39.3), abdomen/pelvis (OR: 16.3; CI: 3.13-80.2), and chest (OR: 49.0; CI: 3.0-794.5). MVI in children more commonly results from firearm-related injury. The mortality rate associated with MVI is profoundly higher than that of the overall pediatric trauma population. These findings underscore the major public health concern of firearm-related injury in children.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
713236
(9R)-6′-Methoxycinchonan-9-amine trihydrochloride, ≥90.0%
C20H25N3O · 3HCl