EMAIL THIS PAGE TO A FRIEND
Oncotarget
GSK3 is required for rapalogs to induce degradation of some oncogenic proteins and to suppress cancer cell growth.
PMID 25797247
Abstract
The single-agent activity of rapalogs (rapamycin and its analogues) in most tumor types has been modest at best. The underlying mechanisms are largely unclear. In this report, we have uncovered a critical role of GSK3 in regulating degradation of some oncogenic proteins induced by rapalogs and cell sensitivity to rapalogs. The basal level of GSK3 activity was positively correlated with cell sensitivity of lung cancer cell lines to rapalogs. GSK3 inhibition antagonized rapamycin's growth inhibitory effects both in vitro and in vivo, while enforced activation of GSK3β sensitized cells to rapamycin. GSK3 inhibition rescued rapamcyin-induced reduction of several oncogenic proteins such as cyclin D1, Mcl-1 and c-Myc, without interfering with the ability of rapamycin to suppress mTORC1 signaling and cap binding. Interestingly, rapamycin induces proteasomal degradation of these oncogenic proteins, as evidenced by their decreased stabilities induced by rapamcyin and rescue of their reduction by proteasomal inhibition. Moreover, acute or short-time rapamycin treatment dissociated not only raptor, but also rictor from mTOR in several tested cell lines, suggesting inhibition of both mTORC1 and mTORC2. Thus, induction of GSK3-dependent degradation of these oncogenic proteins is likely secondary to mTORC2 inhibition; this effect should be critical for rapamycin to exert its anticancer activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA002044
Anti-PSMC4 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA005471
Anti-PSMC4 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105008
Anti-PSMC4, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2105009
Anti-PSMC4, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB4300484
Anti-RPS6 (Ab-235) antibody produced in rabbit, affinity isolated antibody
SAB1300250
Anti-RPS6 (N-term) antibody produced in rabbit, Ig fraction of antiserum, buffered aqueous solution
HPA031153
Anti-RPS6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2107629
Anti-RPS6 antibody produced in rabbit, affinity isolated antibody
GW21987A Anti-SGK (ab1) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
GW21987B Anti-SGK (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1411782
ANTI-SGK antibody produced in mouse, clone 3C4, purified immunoglobulin, buffered aqueous solution
SAB1411783
ANTI-SGK antibody produced in mouse, clone 3G8, purified immunoglobulin, buffered aqueous solution
SAB1411792
ANTI-SGK antibody produced in mouse, clone 1C4, purified immunoglobulin, buffered aqueous solution
S5188
Anti-SGK antibody produced in rabbit, buffered aqueous solution, IgG fraction of antiserum
SAB4502765
Anti-SGK antibody produced in rabbit, affinity isolated antibody
SAB4502764
Anti-SGK, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB2104902
Anti-SGK1, (N-terminal) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
WH0005704M1
Monoclonal Anti-PSMC4 antibody produced in mouse, clone 3G8, purified immunoglobulin, buffered aqueous solution
WH0006194M1
Monoclonal Anti-RPS6 antibody produced in mouse, clone 3H1-F2, purified immunoglobulin, buffered aqueous solution
WH0006446M1
Monoclonal Anti-SGK antibody produced in mouse, clone 4D7-G3, purified immunoglobulin, buffered aqueous solution
SAB1404366
Monoclonal Anti-SGK, (N-terminal) antibody produced in mouse, clone 3E3, purified immunoglobulin, buffered aqueous solution