EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Dodecenylsuccinic anhydride derivatives of gum karaya (Sterculia urens): preparation, characterization, and their antibacterial properties.
PMID 25797306
Abstract
Esterifications of the tree-based gum, gum karaya (GK), using dodecenylsuccinic anhydride (DDSA) were carried out in aqueous solutions. GK was deacetylated using alkali treatment to obtain deacetylated gum karaya (DGK). The DGK and its DDSA derivative were characterized using gel permeation chromatography/multiangle laser light scattering (GPC/MALLS), attenuated total reflectance-Fourier transform infrared spectroscopy (ATR-FTIR), scanning electron microscopy (SEM), proton nuclear magnetic resonance spectroscopy ((1)H NMR), thermogravimetric analysis (TGA), differential scanning calorimetry (DSC) analysis, and rheological studies. The degree of substitution was found to be 10.25% for DGK using (1)H NMR spectroscopy. The critical aggregation concentration of DDSA-DGK was determined using dye solubilization and surface tension methods. The antibacterial activity of the DDSA-DGK derivative was then investigated against Gram-negative Escherichia coli and Pseudomonas aeruginosa and Gram-positive Staphylococcus aureus. The DDSA-DGK derivative has the potential for use as a stabilizing agent in food and nonfood applications. It can also be developed as an antibacterial agent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D221902
(2-Dodecen-1-yl)succinic anhydride, 95%
C16H26O3
44161
(2-Dodecen-1-yl)succinic anhydride, technical
C16H26O3
650455
Cyclohexane, for HPLC, ≥99.9%
C6H12
34855
Cyclohexane, for HPLC, ≥99.7%
C6H12
33117
Cyclohexane, puriss. p.a., ACS reagent, ≥99.5% (GC)
C6H12
179191
Cyclohexane, ACS reagent, ≥99%
C6H12
676861
Cyclohexane, ACS reagent, ≥99%
C6H12
C100307
Cyclohexane, Laboratory Reagent, ≥99.8%
C6H12
443271
Dodecenylsuccinic anhydride, mixture of isomers, technical grade, 90%
C16H26O3
43720
N,N′-Disuccinimidyl carbonate, purum, ≥95.0% (NMR)
C9H8N2O7
746371
Sodium iodide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
INa
793558
Sodium iodide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
INa
439681
Sodium iodide, anhydrous, beads, −10 mesh, 99.999% trace metals basis
INa
409286
Sodium iodide, 99.999% trace metals basis
INa
229911
Sodium iodide, ≥99.99% trace metals basis
INa
S5022
Sodium nitrate, ≥99.0%, plant cell culture tested
NNaO3
229938
Sodium nitrate, 99.995% trace metals basis
NNaO3
15736
Sodium nitrate, analytical standard
NNaO3
S8170
Sodium nitrate, BioXtra, ≥99.0%
NNaO3
347663
Sodium nitrate, beads, 1-2 mm, 98%
NNaO3
71755
Sodium nitrate, BioUltra, ≥99.0% (T)
NNaO3
28-3440
Sodium nitrate, JIS special grade, ≥99.0%
NNaO3
757586
Sodium nitrate, anhydrous, 99.995% trace metals basis
NNaO3
608742
Sodium nitrate-14N, 99.95 atom % 14N
14NNaO3