PloS one
Ion channel expression in the developing enteric nervous system.
PMID 25798587
Abstract
The enteric nervous system arises from neural crest-derived cells (ENCCs) that migrate caudally along the embryonic gut. The expression of ion channels by ENCCs in embryonic mice was investigated using a PCR-based array, RT-PCR and immunohistochemistry. Many ion channels, including chloride, calcium, potassium and sodium channels were already expressed by ENCCs at E11.5. There was an increase in the expression of numerous ion channel genes between E11.5 and E14.5, which coincides with ENCC migration and the first extension of neurites by enteric neurons. Previous studies have shown that a variety of ion channels regulates neurite extension and migration of many cell types. Pharmacological inhibition of a range of chloride or calcium channels had no effect on ENCC migration in cultured explants or neuritogenesis in vitro. The non-selective potassium channel inhibitors, TEA and 4-AP, retarded ENCC migration and neuritogenesis, but only at concentrations that also resulted in cell death. In summary, a large range of ion channels is expressed while ENCCs are colonizing the gut, but we found no evidence that ENCC migration or neuritogenesis requires chloride, calcium or potassium channel activity. Many of the ion channels are likely to be involved in the development of electrical excitability of enteric neurons.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
275875
4-Aminopyridine, ≥99%
C5H6N2
A78403
4-Aminopyridine, 98%
C5H6N2
36687
4-Aminopyridine, PESTANAL®, analytical standard
C5H6N2
N4779
5-Nitro-2-(3-phenylpropylamino)benzoic acid, ≥98%
C16H16N2O4
242381
Benzoic acid, ACS reagent, ≥99.5%
C7H6O2
33045
Benzoic acid, for calorimetrical determination (approx. 26460 J/g)
C7H6O2
06185
Benzoic acid, Standard for quantitative NMR, TraceCERT®
C7H6O2
1055002
Benzoic acid, United States Pharmacopeia (USP) Reference Standard
C7H6O2
33047
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
C7H6O2
18102
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
C7H6O2
12356
Benzoic acid, tested according to Ph.Eur.
C7H6O2
PHR1050
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C7H6O2
12353
Benzoic acid, certified reference material for titrimetry, certified by BAM according to ISO 17025, ≥99.5%
C7H6O2
47849
Benzoic acid, analytical standard
C7H6O2
427608
Benzoic acid, purified by sublimation, ≥99%
C7H6O2
109479
Benzoic acid, ReagentPlus®, 99%
C7H6O2
W213101
Benzoic acid, ≥99.5%, FCC, FG
C7H6O2
W213128
Benzoic acid, natural, ≥99.5%, FCC, FG
C7H6O2
Y0001470
Benzoic acid, European Pharmacopoeia (EP) Reference Standard
C7H6O2
491861
Benzoic acid-12C7, 99.9 atom % 12C
12C7H6O2
B3023
Bumetanide, ≥98%
C17H20N2O5S
B1158000
Bumetanide, European Pharmacopoeia (EP) Reference Standard
C17H20N2O5S
L134
Linopirdine, ≥98% (HPLC)
C26H21N3O
76170
Melting point standard 121-123°C, analytical standard
C7H6O2
73983
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
C7H6O2
8S61336
SS Benzoic Acid, 2000 μg/mL in dichloromethane, analytical standard
C7H6O2
73487
Triethylamine, analytical standard
C6H15N