EMAIL THIS PAGE TO A FRIEND
Marine pollution bulletin
Enterococcus species diversity and molecular characterization of biomarker genes in Enterococcus faecalis in Port Blair Bay, Andaman and Nicobar Islands, India.
PMID 25798888
Abstract
This study was performed to evaluate the abundance and diversity of Enterococcus sp. and the distribution of biomarker genes in Enterococcus faecalis in Port Blair Bay, Andaman and Nicobar Islands. The Enterococcus sp. densities at the seven sampling stations were highly influenced by tidal fluctuations and season. The distributions and diversities of species varied in the inner and outer regions of Port Blair Bay. Among the 1816 total isolates, the occurrence of fecal Enterococcus was high (1.78×10(4) CFU/100 mL) in Phoenix Bay. Moreover, 67.76% of the isolates were identified as Enterococcus, and the most frequently identified species were E. hirae, E. avium and E. faecalis. Assessments of antibiotic resistance and biomarker genes revealed the maximum occurrence in the Aberdeen Bay isolates. The most prevalent biomarker genes observed in the E. faecalis isolates were gelE and asa1, whereas cyl was not found among the isolates. In silico sequence analysis of biomarker genes of E. faecalis also revealed that they are evolutionarily well conserved with those of earlier reports. Further, multivariate analysis distinguished the JB, PB and OS stations from the other stations according to distinctive microbial densities and compositions. In addition, the Shannon-Wiener diversity indices and box-whisker plots further facilitated and supported the multivariate results.