PloS one
Dyslipidemia and chronic inflammation markers are correlated with telomere length shortening in Cushing's syndrome.
PMID 25799396
Abstract
Cushing's syndrome (CS) increases cardiovascular risk (CVR) and adipocytokine imbalance, associated with an increased inflammatory state. Telomere length (TL) shortening is a novel CVR marker, associated with inflammation biomarkers. We hypothesized that inflammatory state and higher CVR in CS might be related to TL shortening, as observed in premature aging. To evaluate relationships between TL, CVR and inflammation markers in CS. In a cross-sectional study, 77 patients with CS (14 males, 59 pituitary-, 17 adrenal- and 1 ectopic-origin; 21 active disease) and 77 age-, gender-, smoking-matched controls were included. Total white blood cell TL was measured by TRF-Southern technique. Clinical data and blood samples were collected (lipids, adrenal function, glucose). Adiponectin, interleukin-6 (IL6) and C-reactive protein (CRP) were available in a subgroup of patients (n=32). Correlations between TL and clinical features were examined and multiple linear regression analysis was performed to investigate potential predictors of TL. Dyslipidemic CS had shorter TL than non-dyslipidemic subjects (7328±1274 vs 7957±1137 bp, p<0.05). After adjustment for age and body mass index, cured and active CS dyslipidemic patients had shorter TL than non-dyslipidemic CS (cured: 7187±1309 vs 7868±1104; active: 7203±1262 vs 8615±1056, respectively, p<0.05). Total cholesterol and triglycerides negatively correlated with TL (r-0.279 and -0.259, respectively, p<0.05), as well as CRP and IL6 (r-0.412 and -0.441, respectively, p<0.05). No difference in TL according the presence of other individual CVR factors (hypertension, diabetes mellitus, obesity) were observed in CS or in the control group. Additional TL shortening was observed in dyslipidemic obese patients who were also hypertensive, compared to those with two or less CVR factors (6956±1280 vs 7860±1180, respectively, p<0.001). Age and dyslipidemia were independent negative predictors of TL. TL is shortened in dyslipidemic CS patients, further worse if hypertension and/or obesity coexist and is negatively correlated with increased inflammation markers. Increased lipids and a "low" grade inflammation may contribute to TL shortening and consequently to premature ageing and increased morbidity in CS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
12209136001
TeloTAGGG Telomere Length Assay