EMAIL THIS PAGE TO A FRIEND
Colloids and surfaces. B, Biointerfaces
Biofunctionalization of microgroove titanium surfaces with an antimicrobial peptide to enhance their bactericidal activity and cytocompatibility.
PMID 25800357
Abstract
A firm peri-implant soft tissue seal is important for the long-term survival of dental implants, which demands the properties of antibacterial and cytocompatibility of the implant surfaces. In this study, GL13K, a cationic antimicrobial peptide, was immobilized onto microgroove surfaces which were 60 μm in width and 10 μm in depth, and the modified surfaces improved both the properties of antibacterial and cytocompatibility. The method of silanization was used to immobilize the antimicrobial peptide GL13K, which was confirmed by X-ray photoelectron spectroscopy (XPS), scanning electron microscopy (SEM), atomic force microscopy (AFM), water contact angle measurement. Then the mechanical stability of the coatings was confirmed by ultrasonication. In vitro antibacterial tests confirmed bactericidal activity against Porphyromonas gingivalis without inhibiting its adhesion. In vitro cytocompatibility tests also confirmed that adhesion at later phase and proliferation of HGFs were greater (P<0.01) on the GL13K-modified microgroove surfaces than on the non-treated microgroove surfaces, and both of them were greater than on the smooth surfaces. The phenomenon of the contact guidance, which is cell growth aligned along the microgrooves, was maintained. Overall, this study developed a promising bi-functional surface that combined the physical and chemical properties to promote cytocompatibility and antibacterial activity simultaneously.