EMAIL THIS PAGE TO A FRIEND
BioMed research international
Use of adipose-derived mesenchymal stem cells in keratoconjunctivitis sicca in a canine model.
PMID 25802852
Abstract
Keratoconjunctivitis sicca (KCS) or dry eye disease (DED) is an immune-mediated multifactorial disease, with high level of prevalence in humans and dogs. Our aim in this study was to investigate the therapeutic effects of allogeneic adipose-derived mesenchymal stromal cells (Ad-MSCs) implanted around the lacrimal glands in 12 dogs (24 eyes) with KCS, which is refractory to current available treatments. Schirmer tear test (STT) and ocular surface integrity were assessed at 0 (before treatment), 3, 6, and 9 months after treatment. Average STT values and all clinical signs showed a statistically significant change (P < 0.001) during the follow-up with reduction in all ocular parameters scored: ocular discharge, conjunctival hyperaemia, and corneal changes, and there were no signs of regression or worsening. Implanted cells were well tolerated and were effective reducing clinical signs of KCS with a sustained effect during the study period. None of the animals showed systemic or local complications during the study. To our knowledge, this is the first time in literature that implantation of allogeneic Ad-MSCs around lacrimal glands has been found as an effective therapeutic alternative to treat dogs with KCS. These results could reinforce a good effective solution to be extrapolated to future studies in human.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1570428
2-Propanol, United States Pharmacopeia (USP) Reference Standard
C3H8O
PHR1072
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
C3H8O
91237
2-Propanol, analytical standard
C3H8O
I7018
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
C10H14N4O2
I5879
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
C10H14N4O2
A9400
7-Aminoactinomycin D, ~97% (HPLC), powder
C62H87N13O16
D1756
Dexamethasone, ≥98% (HPLC), powder
C22H29FO5
D9184
Dexamethasone, meets USP testing specifications
C22H29FO5
D0700000
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
C22H29FO5
1176007
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
C22H29FO5
46165
Dexamethasone, VETRANAL, analytical standard
C22H29FO5
31375
Dexamethasone, tested according to Ph.Eur.
C22H29FO5
PHR1526
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
C22H29FO5
Y0001593
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
C22H29FO5
Y0001177
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
C22H29FO5
I7378
Indomethacin, ≥99% (TLC)
C19H16ClNO4
I8280
Indomethacin, meets USP testing specifications
C19H16ClNO4
1341001
Indomethacin, United States Pharmacopeia (USP) Reference Standard
C19H16ClNO4
I0200000
Indomethacin, European Pharmacopoeia (EP) Reference Standard
C19H16ClNO4
PHR1247
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
C19H16ClNO4
76523
L-Glutamine, certified reference material, TraceCERT®
C5H10N2O3
PHR1125
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
C5H10N2O3
15990
Sodium pyruvate, purum, ≥99.0% (NT)
C3H3NaO3