EMAIL THIS PAGE TO A FRIEND
Plant science : an international journal of experimental plant biology
Ectopic expression of a stress-inducible glycosyltransferase from saffron enhances salt and oxidative stress tolerance in Arabidopsis while alters anchor root formation.
PMID 25804810
Abstract
Glycosyltransferases play diverse roles in cellular metabolism by modifying the activities of regulatory metabolites. Three stress-regulated UDP-glucosyltransferase-encoding genes have been isolated from the stigmas of saffron, UGT85U1, UGT85U2 and UGT85V1, which belong to the UGT85 family that includes members associated with stress responses and cell cycle regulation. Arabidopsis constitutively expressing UGT85U1 exhibited and increased anchor root development. No differences were observed in the timing of root emergence, in leaf, stem and flower morphology or flowering time. However, salt and oxidative stress tolerance was enhanced in these plants. Levels of glycosylated compounds were measured in these plants and showed changes in the composition of several indole-derivatives. Moreover, auxin levels in the roots were higher compared to wild type. The expression of several key genes related to root development and auxin homeostasis, including CDKB2.1, CDKB2.2, PIN2, 3 and 4; TIR1, SHR, and CYCD6, were differentially regulated with an increase of expression level of SHR, CYCD6, CDKB2.1 and PIN2. The obtained results showed that UGT85U1 takes part in root growth regulation via auxin signal alteration and the modified expression of cell cycle-related genes, resulting in significantly improved survival during oxidative and salt stress treatments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36333
3-Hydroxybenzoic acid, analytical standard
C7H6O3
H20008
3-Hydroxybenzoic acid, ReagentPlus®, 99%
C7H6O3
45533
3-Indoleacetic acid, PESTANAL®, analytical standard
C10H9NO2
246573
6,7-Dihydroxycoumarin, 98%
C9H6O4
72609
Coumarin, certified reference material, TraceCERT®
C9H6O2
C4261
Coumarin, ≥99% (HPLC)
C9H6O2
W526509
Coumarin, ≥98%
C9H6O2
01260595
Coumarin, primary pharmaceutical reference standard
C9H6O2
Y0000438
Coumarin, European Pharmacopoeia (EP) Reference Standard
C9H6O2
D7802
Daidzein, ≥98%, synthetic
C15H10O4
16587
Daidzein, analytical standard
C15H10O4
Y0001611
Esculetin, European Pharmacopoeia (EP) Reference Standard
C9H6O4
92136
Genistein, analytical standard
C15H10O5
S5922
Salicylic acid, BioXtra, ≥99.0%
C7H6O3
1609002
Salicylic acid, United States Pharmacopeia (USP) Reference Standard
C7H6O3
84211
Salicylic acid, tested according to Ph.Eur.
C7H6O3
PHR1013
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
C7H6O3
105910
Salicylic acid, ReagentPlus®, ≥99%
C7H6O3
W398500
Salicylic acid, ≥99%, FG
C7H6O3
52341
Salicylic acid, certified reference material, TraceCERT®
C7H6O3
S0200000
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
C7H6O3
S-019
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material
C7H6O3
49508
Sinapic acid, matrix substance for MALDI-MS, ≥99.5%, Ultra pure
C11H12O5
85429
Sinapic acid, matrix substance for MALDI-MS, ≥99.0% (T)
C11H12O5
96340
trans-Cinnamic acid, purum, ≥99.0% (T)
C9H8O2
C80857
trans-Cinnamic acid, ≥99%
C9H8O2
W228818
trans-Cinnamic acid, ≥99%, FG
C9H8O2
97013
trans-Cinnamic acid, analytical standard
C9H8O2
W228826
trans-Cinnamic acid, natural, ≥99%, FCC, FG
C9H8O2
133760
trans-Cinnamic acid, 97%
C9H8O2