EMAIL THIS PAGE TO A FRIEND
Clinica chimica acta; international journal of clinical chemistry
Comparing the alpha-galactosidase A biochemical properties from healthy individuals and Fabry disease patients.
PMID 25804996
Abstract
Due to the importance and the difficulty still present in determining the biochemical diagnosis of Fabry disease (FD), the aim of this study was to establish and compare the biochemical and kinetic properties of alpha-galactosidase A (GLA) in dried blood spots (DBS), plasma and leukocyte samples of FD patients and healthy subjects to evaluate the possible use of these parameters as an auxiliary tool in the diagnosis of this disease. GLA activity in DBS, plasma and leukocyte samples from Fabry disease patients and healthy subjects was compared and characterized in terms of optimal pH, Km and Vmax and heat stability. A difference was observed between the Km and Vmax of FD patients and healthy controls using DBS, plasma and leukocyte samples. In leukocytes, pre-incubation at 50°C for 60 min was effective to differentiate FD patients from healthy controls. These results can be used as an auxiliary method to the FD diagnosis, especially in cases of patients whose GLA activity is within normal range.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2795
N-Acetyl-D-galactosamine, ~98%
C8H15NO6