EMAIL THIS PAGE TO A FRIEND
Clinical cancer research : an official journal of the American Association for Cancer Research
Biologic Activity of Autologous, Granulocyte-Macrophage Colony-Stimulating Factor Secreting Alveolar Soft-Part Sarcoma and Clear Cell Sarcoma Vaccines.
PMID 25805798
Abstract
Alveolar soft-part sarcoma (ASPS) and clear cell sarcoma (CCS) are rare mesenchymal malignancies driven by chromosomal translocations that activate members of the microphthalmia transcription factor (MITF) family. However, in contrast to malignant melanoma, little is known about their immunogenicity. To learn more about the host response to ASPS and CCS, we conducted a phase I clinical trial of vaccination with irradiated, autologous sarcoma cells engineered by adenoviral-mediated gene transfer to secrete granulocyte-macrophage colony-stimulating factor (GM-CSF). Metastatic tumors from ASPS and CCS patients were resected, processed to single-cell suspensions, transduced with a replication-defective adenoviral vector encoding GM-CSF, and irradiated. Immunizations were administered subcutaneously and intradermally weekly three times and then every other week. Vaccines were successfully manufactured for 11 of the 12 enrolled patients. Eleven subjects received from three to 13 immunizations. Toxicities were restricted to grade 1-2 skin reactions at inoculation sites. Vaccination elicited local dendritic cell infiltrates and stimulated T cell-mediated delayed-type hypersensitivity reactions to irradiated, autologous tumor cells. Antibody responses to tissue-type plasminogen activator (tTPA) and angiopoietins-1/2 were detected. Tumor biopsies showed programmed death-1 (PD-1)-positive CD8(+) T cells in association with PD ligand-1 (PD-L1)-expressing sarcoma cells. No tumor regressions were observed. Vaccination with irradiated, GM-CSF-secreting autologous sarcoma cell vaccines is feasible, safe, and biologically active. Concurrent targeting of angiogenic cytokines and antagonism of the PD-1-negative regulatory pathway might intensify immune-mediated tumor destruction.