EMAIL THIS PAGE TO A FRIEND
Kidney international
Gluten exacerbates IgA nephropathy in humanized mice through gliadin-CD89 interaction.
PMID 25807036
Abstract
IgA1 complexes containing deglycosylated IgA1, IgG autoantibodies, and a soluble form of the IgA receptor (sCD89), are hallmarks of IgA nephropathy (IgAN). Food antigens, notably gluten, are associated with increased mucosal response and IgAN onset, but their implication in the pathology remains unknown. Here, an IgAN mouse model expressing human IgA1 and CD89 was used to examine the role of gluten in IgAN. Mice were given a gluten-free diet for three generations to produce gluten sensitivity, and then challenged for 30 days with a gluten diet. A gluten-free diet resulted in a decrease of mesangial IgA1 deposits, transferrin 1 receptor, and transglutaminase 2 expression, as well as hematuria. Mice on a gluten-free diet lacked IgA1-sCD89 complexes in serum and kidney eluates. Disease severity depended on gluten and CD89, as shown by reappearance of IgAN features in mice on a gluten diet and by direct binding of the gluten-subcomponent gliadin to sCD89. A gluten diet exacerbated intestinal IgA1 secretion, inflammation, and villous atrophy, and increased serum IgA1 anti-gliadin antibodies, which correlated with proteinuria in mice and patients. Moreover, early treatment of humanized mice with a gluten-free diet prevented mesangial IgA1 deposits and hematuria. Thus, gliadin-CD89 interaction may aggravate IgAN development through induction of IgA1-sCD89 complex formation and a mucosal immune response. Hence, early-stage treatment with a gluten-free diet could be beneficial to prevent disease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N9389
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
C6H4NNa2O6P · 6H2O
N2765
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
C6H4NNa2O6P · 6H2O
N2640
4-Nitrophenyl phosphate disodium salt hexahydrate, tablet
C6H4NNa2O6P · 6H2O
N4645
4-Nitrophenyl phosphate disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%
C6H4NNa2O6P · 6H2O
71768
4-Nitrophenyl phosphate disodium salt hexahydrate, for enzyme immunoassay, ≥99.0% (enzymatic)
C6H4NNa2O6P · 6H2O
M0945 Anti-Maspin antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB1306340
ANTI-PAI2 (CENTER) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1406218
Anti-SERPINB2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA015480
Anti-SERPINB2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104454
Anti-SERPINB2 antibody produced in rabbit, affinity isolated antibody
SAB1410870
Anti-SERPINB2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA020136
Anti-SERPINB5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab3
HPA019025
Anti-SERPINB5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA019132
Anti-SERPINB5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
SAB4502191
Anti-SERPINB5 antibody produced in rabbit, affinity isolated antibody
HPA024200
Anti-SERPINB7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1401435
Anti-SERPINB7 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4503460
Anti-SERPINB7 antibody produced in rabbit, affinity isolated antibody
SAB1400210
Anti-SERPINB8 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2500930
Anti-SERPINB9 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA030067
Anti-SERPINB9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502192
Anti-SERPINB9 antibody produced in rabbit, affinity isolated antibody
WH0005055M8
Monoclonal Anti-SERPINB2 antibody produced in mouse, clone 3A9, purified immunoglobulin, buffered aqueous solution
SAB1402869
Monoclonal Anti-SERPINB8 antibody produced in mouse, clone 1D10, ascites fluid
SAB1402309
Monoclonal Anti-SERPINB9, (C-terminal) antibody produced in mouse, clone 1F5, purified immunoglobulin, buffered aqueous solution
P4744
Phosphatase substrate, powder
C6H4NNa2O6P · 6H2O
S0942
Phosphatase substrate, 5 mg tablets
C6H4NNa2O6P · 6H2O
P5994
Phosphatase substrate, 40 mg tablets
C6H4NNa2O6P · 6H2O
P5744
Phosphatase substrate, 40 mg capsules
C6H4NNa2O6P · 6H2O
P5869
Phosphatase substrate, 100 mg capsules
C6H4NNa2O6P · 6H2O