EMAIL THIS PAGE TO A FRIEND
Cancer research
Decoy Receptor DcR1 Is Induced in a p50/Bcl3-Dependent Manner and Attenuates the Efficacy of Temozolomide.
PMID 25808868
Abstract
Temozolomide is used widely to treat malignant glioma, but the overall response to this agent is generally poor. Resistance to DNA-damaging drugs such as temozolomide has been related to the induction of antiapoptotic proteins. Specifically, the transcription factor NF-κB has been suggested to participate in promoting the survival of cells exposed to chemotherapy. To identify factors that modulate cytotoxicity in the setting of DNA damage, we used an unbiased strategy to examine the NF-κB-dependent expression profile induced by temozolomide. By this route, we defined the decoy receptor DcR1 as a temozolomide response gene induced by a mechanism relying upon p50/NF-κB1. A conserved NF-κB-binding sequence (κB-site) was identified in the proximal promoter and was demonstrated to be required for DcR1 induction by temozolomide. Loss-of-function and gain-of-function studies reveal that the atypical IκB protein, Bcl3, is also required for induction of DcR1 by temozolomide. Mechanistically, DcR1 attenuates temozolomide efficacy by blunting activation of the Fas receptor pathway in p53(+/+) glioma cells. Intracranial xenograft studies show that DcR1 depletion in glioma cells enhances the efficacy of temozolomide. Taken together, our results show how DcR1 upregulation mediates temozolomide resistance and provide a rationale for DcR1 targeting as a strategy to sensitize gliomas to this widely used chemotherapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1405511
Anti-BCL3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401028
Anti-BCL3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2702024
Anti-BCL3 antibody produced in rabbit
SAB3501028
Anti-BCL3 antibody produced in rabbit, affinity isolated antibody
HPA003928
Anti-CDC37 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100393
Anti-CEACAM16 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2100394
Anti-CEACAM16 (ab2) antibody produced in rabbit, affinity isolated antibody
HPA045075
Anti-CEACAM16 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1407011
Anti-DCTN2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2100543
Anti-DCTN2 antibody produced in rabbit, affinity isolated antibody
HPA039715
Anti-DCTN2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA040040
Anti-DCTN2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2501426
Anti-LSP1 (C-terminal) antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501581
Anti-LSP1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA019693
Anti-LSP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101400
Anti-LSP1 antibody produced in rabbit, affinity isolated antibody
SAB1410762
Anti-LSP1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4500124
Anti-p50 Dynamitin, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1400218
Anti-POLD2 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
HPA026745
Anti-POLD2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101838
Anti-POLD2 antibody produced in rabbit, affinity isolated antibody
SAB1410988
Anti-POLD2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
WH0011140M1
Monoclonal Anti-CDC37 antibody produced in mouse, clone 3C7, purified immunoglobulin, buffered aqueous solution
WH0010540M1
Monoclonal Anti-DCTN2 antibody produced in mouse, clone 2G7, purified immunoglobulin, buffered aqueous solution
SAB1400155
Monoclonal Anti-LSP1 antibody produced in mouse, clone 1C4, purified immunoglobulin, buffered aqueous solution
T2577
Temozolomide, ≥98% (HPLC)
C6H6N6O2