EMAIL THIS PAGE TO A FRIEND
Nature communications
Orphan receptor IL-17RD regulates Toll-like receptor signalling via SEFIR/TIR interactions.
PMID 25808990
Abstract
Receptor families of the innate immune response engage in 'cross-talk' to tailor optimal immune responses against invading pathogens. However, these responses are subject to multiple levels of regulation to keep in check aberrant inflammatory signals. Here, we describe a role for the orphan receptor interleukin-17 receptor D (IL-17RD) in negatively regulating Toll-like receptor (TLR)-induced responses. Deficiency of IL-17RD expression in cells leads to enhanced pro-inflammatory signalling and gene expression in response to TLR stimulation, and Il17rd(-/-) mice are more susceptible to TLR-induced septic shock. We demonstrate that the intracellular Sef/IL-17R (SEFIR) domain of IL-17RD targets TIR adaptor proteins to inhibit TLR downstream signalling thus revealing a paradigm involving cross-regulation of members of the IL-17R and TLR families.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4501565
Anti-IRF-3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1406043
Anti-IRF3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA004895
Anti-IRF3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV31992
Anti-IRF3 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
AV38061
Anti-IRF3 antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
SAB3500280
Anti-IRF3 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4501563
Anti-IRF3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501564
Anti-IRF3, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1406553
Anti-TRAF6 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA020599
Anti-TRAF6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
HPA019805
Anti-TRAF6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
SAB3500377
Anti-TRAF6 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
RAB0119
Human IP-10 / CXCL10 ELISA Kit, for serum, plasma, cell culture supernatant and urine
EMU041271 MISSION® esiRNA, esiRNA targeting mouse Il17rd (esiRNA1)
EHU079801 MISSION® esiRNA, esiRNA human IL17RD (esiRNA1)
SAB4100036
Monoclonal Anti-IRF3 antibody produced in mouse, culture supernatant
SAB4100037
Monoclonal Anti-IRF3 antibody produced in mouse, culture supernatant
SAB1402824
Monoclonal Anti-IRF3 antibody produced in mouse, clone 3C8, purified immunoglobulin, buffered aqueous solution
WH0007189M2
Monoclonal Anti-TRAF6 antibody produced in mouse, clone 1B2, purified immunoglobulin, buffered aqueous solution
RAB0120
Mouse CRG-2 / CXCL10 ELISA Kit, for serum, plasma and cell culture supernatant