EMAIL THIS PAGE TO A FRIEND
Phytotherapy research : PTR
Castalagin Exerts Inhibitory Effects on Osteoclastogenesis Through Blocking a Broad Range of Signaling Pathways with Low Cytotoxicity.
PMID 25809147
Abstract
Castalagin is a rare plant polyphenol that is classified as a hydrolyzable tannin. Although it has antioxidant, antitumorigenic, and leishmanicidal effects, the utility of castalagin against bone diseases remain to be elucidated. Here, we investigated the effects of castalagin on the differentiation of osteoclasts (OCLs), multinucleated bone-resorbing cells. After stimulation with receptor activator of nuclear factor kappa-B ligand (RANKL), the formation of OCLs from bone marrow-derived macrophages was significantly inhibited by castalagin even at 1 μM. However, castalagin displayed little cytotoxicity at a higher concentration of 50 μM. The effects of castalagin on intracellular signaling during OCL differentiation showed that castalagin suppresses RANKL-stimulated phosphorylation of major signaling pathways including protein kinase B (Akt), extracellular signal-regulated kinase, Jun N-terminal kinase, p38 mitogen-activated protein kinases, and inhibitor of nuclear factor kappa B alpha. Moreover, following castalagin treatment, the protein levels of nuclear factor of activated T-cells, cytoplasmic 1, a master regulator for OCL differentiation, and NF-κB were decreased. Thus, castalagin exerts inhibitory effects on osteoclastogenesis through blockage of a broad range of signaling pathways, but has low cytotoxicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2500203
Anti-Cathepsin K antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501977
Anti-CSF1R antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2501384
Anti-CSF1R antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA012323
Anti-CSF1R antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4500500
Anti-CSFR antibody produced in rabbit, affinity isolated antibody
SAB4500499
Anti-CSFR, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1405680
Anti-CTSK antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410267
Anti-CTSK antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA027728
Anti-TNFRSF11A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
C8243
Monoclonal Anti-Cathepsin K antibody produced in mouse, ~2 mg/mL, clone CL124-1H6, purified immunoglobulin, buffered aqueous solution
WH0001436M1
Monoclonal Anti-CSF1R antibody produced in mouse, clone 1G4, purified immunoglobulin, buffered aqueous solution
SAB1403703
Monoclonal Anti-CSF1R antibody produced in mouse, clone 3G12, purified immunoglobulin, buffered aqueous solution
WH0001513M1
Monoclonal Anti-CTSK antibody produced in mouse, clone 2F1, purified immunoglobulin, buffered aqueous solution
SAB1403714
Monoclonal Anti-CTSK antibody produced in mouse, clone 1B11, purified immunoglobulin, buffered aqueous solution