EMAIL THIS PAGE TO A FRIEND
Cellular and molecular life sciences : CMLS
Influenza infection induces host DNA damage and dynamic DNA damage responses during tissue regeneration.
PMID 25809161
Abstract
Influenza viruses account for significant morbidity worldwide. Inflammatory responses, including excessive generation of reactive oxygen and nitrogen species (RONS), mediate lung injury in severe influenza infections. However, the molecular basis of inflammation-induced lung damage is not fully understood. Here, we studied influenza H1N1 infected cells in vitro, as well as H1N1 infected mice, and we monitored molecular and cellular responses over the course of 2 weeks in vivo. We show that influenza induces DNA damage to both, when cells are directly exposed to virus in vitro (measured using the comet assay) and also when cells are exposed to virus in vivo (estimated via γH2AX foci). We show that DNA damage, as well as responses to DNA damage persist in vivo until long after virus has been cleared, at times when there are inflammation associated RONS (measured by xanthine oxidase activity and oxidative products). The frequency of lung epithelial and immune cells with increased γH2AX foci is elevated in vivo, especially for dividing cells (Ki-67-positive) exposed to oxidative stress during tissue regeneration. Additionally, we observed a significant increase in apoptotic cells as well as increased levels of DNA double strand break (DSB) repair proteins Ku70, Ku86 and Rad51 during the regenerative phase. In conclusion, results show that influenza induces DNA damage both in vitro and in vivo, and that DNA damage responses are activated, raising the possibility that DNA repair capacity may be a determining factor for tissue recovery and disease outcome.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV34116
Anti-G22P1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV34117
Anti-G22P1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB4300514
Anti-H2AFX (Ab-139) antibody produced in rabbit, affinity isolated antibody
SAB4501369
Anti-Histone H2AX , C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB1306517
ANTI-KU70 (CENTER) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
K4763 Anti-Ku70 antibody produced in rabbit, affinity isolated antibody, ammonium sulfate suspension
SAB4501057
Anti-KU70, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503096
Anti-KU70, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4503095
Anti-KU80, C-Terminal antibody produced in rabbit, affinity isolated antibody
AV34121
Anti-XRCC5 antibody produced in rabbit, IgG fraction of antiserum
HPA025813
Anti-XRCC5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503098
Anti-XRCC5 antibody produced in rabbit, affinity isolated antibody
SAB4503097
Anti-XRCC5, C-Terminal antibody produced in rabbit, affinity isolated antibody
SAB4501058
Anti-XRCC6, C-Terminal antibody produced in rabbit, affinity isolated antibody
PLA0294
Goat anti-H2AX Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
WH0003014M5 Monoclonal Anti-H2AFX antibody produced in mouse, clone 3F4, purified immunoglobulin, buffered aqueous solution
SAB1403902 Monoclonal Anti-H2AFX, (N-terminal) antibody produced in mouse, clone 2H5, purified immunoglobulin, buffered aqueous solution
WH0002547M1
Monoclonal Anti-XRCC6 antibody produced in mouse, clone 4C2-1A6, purified immunoglobulin, buffered aqueous solution
SAB1403847
Monoclonal Anti-XRCC6 antibody produced in mouse, clone 3D12, purified immunoglobulin, buffered aqueous solution
PLA0023
Rabbit anti-H2AX Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
PLA0263
Rabbit anti-Ku70 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
PLA0264
Rabbit anti-Ku80 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.