EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Vibsanin B preferentially targets HSP90β, inhibits interstitial leukocyte migration, and ameliorates experimental autoimmune encephalomyelitis.
PMID 25810397
Abstract
Interstitial leukocyte migration plays a critical role in inflammation and offers a therapeutic target for treating inflammation-associated diseases such as multiple sclerosis. Identifying small molecules to inhibit undesired leukocyte migration provides promise for the treatment of these disorders. In this study, we identified vibsanin B, a novel macrocyclic diterpenoid isolated from Viburnum odoratissimum Ker-Gawl, that inhibited zebrafish interstitial leukocyte migration using a transgenic zebrafish line (TG:zlyz-enhanced GFP). We found that vibsanin B preferentially binds to heat shock protein (HSP)90β. At the molecular level, inactivation of HSP90 can mimic vibsanin B's effect of inhibiting interstitial leukocyte migration. Furthermore, we demonstrated that vibsanin B ameliorates experimental autoimmune encephalomyelitis in mice with pathological manifestation of decreased leukocyte infiltration into their CNS. In summary, vibsanin B is a novel lead compound that preferentially targets HSP90β and inhibits interstitial leukocyte migration, offering a promising drug lead for treating inflammation-associated diseases.