EMAIL THIS PAGE TO A FRIEND
Journal of virology
Borna disease virus phosphoprotein modulates epigenetic signaling in neurons to control viral replication.
PMID 25810554
Abstract
Understanding the modalities of interaction of neurotropic viruses with their target cells represents a major challenge that may improve our knowledge of many human neurological disorders for which viral origin is suspected. Borna disease virus (BDV) represents an ideal model to analyze the molecular mechanisms of viral persistence in neurons and its consequences for neuronal homeostasis. It is now established that BDV ensures its long-term maintenance in infected cells through a stable interaction of viral components with the host cell chromatin, in particular, with core histones. This has led to our hypothesis that such an interaction may trigger epigenetic changes in the host cell. Here, we focused on histone acetylation, which plays key roles in epigenetic regulation of gene expression, notably for neurons. We performed a comparative analysis of histone acetylation patterns of neurons infected or not infected by BDV, which revealed that infection decreases histone acetylation on selected lysine residues. We showed that the BDV phosphoprotein (P) is responsible for these perturbations, even when it is expressed alone independently of the viral context, and that this action depends on its phosphorylation by protein kinase C. We also demonstrated that BDV P inhibits cellular histone acetyltransferase activities. Finally, by pharmacologically manipulating cellular acetylation levels, we observed that inhibiting cellular acetyl transferases reduces viral replication in cell culture. Our findings reveal that manipulation of cellular epigenetics by BDV could be a means to modulate viral replication and thus illustrate a fascinating example of virus-host cell interaction. Persistent DNA viruses often subvert the mechanisms that regulate cellular chromatin dynamics, thereby benefitting from the resulting epigenetic changes to create a favorable milieu for their latent and persistent states. Here, we reasoned that Borna disease virus (BDV), the only RNA virus known to durably persist in the nucleus of infected cells, notably neurons, might employ a similar mechanism. In this study, we uncovered a novel modality of virus-cell interaction in which BDV phosphoprotein inhibits cellular histone acetylation by interfering with histone acetyltransferase activities. Manipulation of cellular histone acetylation is accompanied by a modulation of viral replication, revealing a perfect adaptation of this "ancient" virus to its host that may favor neuronal persistence and limit cellular damage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1409834
Anti-CCDC6 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA019049
Anti-CCDC6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
HPA019051
Anti-CCDC6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
SAB2107667
Anti-CCDC6 antibody produced in rabbit, affinity isolated antibody
SAB1411455
Anti-CCDC6 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1410334
Anti-EMP3 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4300619
Anti-HIST2H2BE antibody produced in rabbit, affinity isolated antibody
SAB2107598
Anti-HIST2H4 antibody produced in rabbit, affinity isolated antibody
SAB1300570
Anti-HRH4 (C-term) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2701955
Anti-HRH4 (N-terminal) antibody produced in rabbit
HPA035009
Anti-HRH4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502598
Anti-HRH4 antibody produced in rabbit, affinity isolated antibody
SAB1401658
Anti-ICOS antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1306778
ANTI-ICOS(C-TERMINAL) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2102105
Anti-SELS antibody produced in rabbit, affinity isolated antibody
SAB1411081
Anti-SELS antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1407701
Anti-SELS, (N-terminal) antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA010025
Anti-VIMP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
WH0008030M3
Monoclonal Anti-CCDC6 antibody produced in mouse, clone 5D11, purified immunoglobulin, buffered aqueous solution
WH0002014M1
Monoclonal Anti-EMP3 antibody produced in mouse, clone 3D4, purified immunoglobulin, buffered aqueous solution
SAB1403769
Monoclonal Anti-EMP3 antibody produced in mouse, clone 2C4, purified immunoglobulin, buffered aqueous solution
SAB1402418
Monoclonal Anti-HIST2H2BE, (C-terminal) antibody produced in mouse, clone 4G6, purified immunoglobulin, buffered aqueous solution
WH0029851M1 Monoclonal Anti-ICOS antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
SAB1412835
Monoclonal Anti-ICOS antibody produced in mouse, clone 3G4, purified immunoglobulin, 0.5 mg/mL