PloS one
Pharmacokinetics and Metabolism of 4R-Cembranoid.
PMID 25811857
Abstract
4R-cembranoid (4R) is a natural cyclic diterpenoid found in tobacco leaves that displays neuroprotective activity. 4R protects against NMDA, paraoxon (POX), and diisopropylfluorophosphate (DFP) damage in rat hippocampal slices and against DFP in rats in vivo. The purpose of this study was to examine the metabolism and pharmacokinetics of 4R as part of its preclinical development as a neuroprotective drug. 10 µM 4R was found to be very stable in plasma for up to 1 hr incubation. 4R metabolism in human microsomes was faster than in the rat. Ten metabolites of 4R were detected in the microsomal samples; 6 dihydroxy and 4 monohydroxy forms of 4R. Male rats received a single dose of 4R at 6 mg/kg i.v., i.m., or s.c. The i.v. group had the highest plasma concentration of 1017 ng/mL. The t1/2 was 36 min and reached the brain within 10 min. The brain peak concentration was 6516 ng/g. The peak plasma concentration in the i.m. group was 163 ng/mL compared to 138 ng/mL in the s.c. group. The t1/2 of 4R after i.m. and s.c. administration was approximately 1.5 hr. The brain peak concentration was 329 ng/g in the i.m. group and 323 ng/g for the s.c. group. The brain to plasma ratio in the i.v. group was 6.4, reached 10 min after dose, whereas in the i.m. and s.c. groups was 2.49 and 2.48, respectively, at 90 min after dose. Our data show that 4R crosses the BBB and concentrates in the brain where it exerts its neuroprotective effect.